Customer Education
Aldrich Materials Science Presents a Technical Webinar – High-capacity drug-delivery systems for cancer therapy based on poly(2-oxazoline)s
Webinar Title: High-capacity drug-delivery systems for cancer therapy based on poly(2-oxazoline)s
You can watch the recorded version of the webinar by clicking the button above.
 What Does it Cover?
Poly(2-oxazoline) (POx) is a very promising candidate for use in polymer therapeutics. Amphiphilic POx triblock copolymers can be used as high capacity drug delivery systems for hydrophobic drugs, like paclitaxel, and exhibit synergistic effects for the delivery of multiple chemotherapeutics.  In this talk, Professor Jordan will highlight the unparalleled high drug-loading capacities of POx systems, the polymer structural variability, and outline the consequences of the drug loading on the drug delivery system morphology.
 What Will You Learn?
  • Properties and advantages of polyoxazolines
  • Methods for drug formulation with polyoxazolines
  • Drug loading in polyoxazolines-based drug delivery systems
  • Application of polyoxazoline drug delivery systems (cancer)
 Who Should Attend?
  • Pharmaceutical Scientists
  • Biomedical Engineers
  • Medicinal Chemists
  • Polymer Chemists
  • Drug Delivery Researchers
Speaker Bio
Prof. Rainer Jordan
Chair of Macromolecular Chemistry, School of Science
Technische Universität Dresden, Dresden, Germany
Dr. Rainer Jordan studied chemistry at the University of Mainz, Germany and as an IAS fellow with Prof. T. Saegusa at Kyoto University, Japan. He joined Prof. K.K. Unger’s group in Mainz and worked as a PROCOPE fellow at C.N.R.S. Paris. In 1996, he obtained his doctoral degree in Chemistry with Prof. Unger (University of Mainz). After a postdoctorate with Prof. Ulman at Polytechnic University in Brooklyn, NY, USA, he was appointed as Assistant Professor. He returned to Germany and joined the group of Prof. O. Nuyken at the Technische Universität München for Habilitation. Since 2008/2009 he is Full Professor, Chair of Macromolecular Chemistry at the Technische Universität Dresden, Germany. His research interests include polymer chemistry, surface chemistry, self-assembly, biomimetic systems, nanoscience, and nanomedicine.
For more information, email