Webinar: PAH Analysis with GC Columns - Most Popular Columns
PAH Analysis webinar
 Watch Now
 Webinar Overview
In 1984, the US EPA promulgated Method 610 for the analysis of Polynuclear Aromatic Hydrocarbons. The original method allows for the use of HPLC and GC techniques, but specifically states the GC technique “does not adequately resolve the following four pairs of compounds: anthracene and phenanthrene; chrysene and benzo[a]anthracene; benzo[b]fluoranthene and benzo[k]fluoranthene; and dibenzo[a,h] anthracene and indeno (1,2,3-cd)pyrene”. We recently launched SLB-ILPAH columns that allow for better resolution of the isomer sets originally called out in Method 610. These new columns provide better resolution and are stable up to 350˚C. This webinar will cover the results of comparison studies of the new columns with more conventional columns as well as the chemistry behind them.
 Who Should Watch
  • Anyone performing PAH analysis
 You Will Learn About:
  • Two new specialty columns that have been developed for the analysis of PAHs
    - SLB-ILPAH (ideal for GC/FID/ECD or other non-MS detectors)
    - SLB-PAHms (ideal for GC/MS)
Speaker Bio
Lisa McCombie
Global Product Manager
Gas Chromatography and Specialty Carbon Adsorbents Advanced Analytical
Lisa McCombie is the Global Product Manager for Gas Chromatography and Specialty Carbon Adsorbents. Prior to her current role, Lisa spent six years in various analytical chemistry roles including formulations chemist for analytical reference materials, global product manager for proficiency testing, and global marketing manager for air monitoring. She holds a bachelor’s degree in Chemistry and an MBA.