Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Enzyme Explorer
Product List
Complement Factor Proteins
Related Proteins
Deficient Sera
Complement Sera
Related Antibodies
Assay Reagents
Back to Complement Proteins
Plasma & Blood Protein Resource
Complement Factor Proteins  
Product Name Product # 
Complement C3 from human serum, approx.95% (SDS-PAGE), Lyophilized powder C0651
Complement component C1q from human serum, Lyophilized powder C0660
Complement C1r from human serum, 0.1 mg/mL protein in 20 mM Tris-HCl, 154 mM NaCl, 1 mM EDTA, and 0.03 mM p-nitrophenyl-p¢-guanidinobenzoate, pH 7.4, approx.90% (nonreduced SDS-urea gel) C2162
Complement C1 from human serum, approx.90% (nonreduced SDS-urea gel) C2660
Complement C7 from human serum, 1 mg/mL in 10 mM sodium phosphate, 150 mM sodium chloride, pH 7.2, 60% (SDS-PAGE), >200,000 C7H50 units/mg protein C2787
Complement C3 from human serum, minimum50 C3H50 units/mg (using C3 deficient serum) C2910
Complement C5 from human serum, 1 mg/mL in 10 mM sodium phosphate, 150 mM sodium chloride, pH 7.2, 75% (SDS-PAGE), >300,000 C5H50 units/mg protein C3160
Complement C6 from human serum, 1 mg/mL in 10 mM sodium phosphate, 150 mM sodium chloride, pH 7.2, 80% (SDS-PAGE), >300,000 C6H50 units/mg protein C3285
Complement C8 from human serum, 1 mg/mL in 15 mM sodium phosphate, 150 mM NaCl, pH 7.2, suitable for radioiodination, minimum85% (SDS-PAGE), minimum125,000 C8H50 units/mg protein (using C8 deficient serum) C3535
Complement C9 from human serum, 1 mg/mL in 15 mM sodium phosphate, 135 mM NaCl, pH 7.4, minimum70,000 C9H50 units/mg (using C9 deficient serum) C3660
Complement factor D from human plasma, 100 µg/mL in PBS, pH 7.2, >90% (SDS-PAGE) C5688
Complement C5a human, approx.95% (SDS-PAGE and HPLC), recombinant, expressed in Escherichia coli, Lyophilized powder C5788
Complement factor H from human plasma, 1 mg/mL in PBS, pH 7.2, >90% (SDS-PAGE) C5813
Complement factor I from human plasma, 1 mg/mL in PBS, pH 7.2, >90% (SDS-PAGE) C5938
Complement C4 from human serum, 1 mg/mL in 25 mM sodium phosphate, 100 mM NaCl, pH 7.2, containing 0.02% sodium azide., minimum300,000 C4H50 units/mg protein C8195
Related Proteins back to top
Product Name Product # 
Esterase inhibitor C-1 from human plasma, minimum90% (SDS-PAGE) C2412
Cobra venom anti-complementary protein Naja naja kaouthia, Lyophilized powder C8406
Deficient Sera back to top
Product Name Product # 
Complement C2 deficient serum human C0913
Complement C4 deficient serum from guinea pig C1038
Complement C5 deficient serum human C1163
Complement C6 deficient serum human C1288
Complement C7 deficient serum human C1413
Complement C8 deficient serum human C1538
Complement C9 deficient serum human C1663
Complement C3 deficient serum human C8788
Complement Sera back to top
Product Name Product # 
Complement sera Complement Serum Standard human, Aqueous solution C9473
Complement sera from guinea pig, Lyophilized powder S1639
Complement sera human, Lyophilized powder S1764
Complement sera from mouse, Lyophilized powder S3269
Complement sera from rat, Lyophilized powder S3394
Complement sera from rabbit, Lyophilized powder S7764
Related Antibodies back to top
Product Name Product # 
Anti-Complement C4 antibody produced in rabbit, Lyophilized powder, IgG fraction of antiserum C3402
Anti-Complement C1q antibody produced in goat, Lyophilized powder, Fractionated antiserum C3900
Anti-Complement C3c antibody produced in rabbit, IgG fraction of antiserum, Lyophilized powder C6025
Anti-Complement C3 antibody produced in goat, Fractionated antiserum, Lyophilized powder C7761
Anti-Sheep Red Blood Cell Stroma antibody produced in rabbit, IgG fraction of antiserum, Lyophilized powder S8014
Assay Reagents back to top
Product Name Product # 
Antibody Sensitized Sheep Erythrocytes, 1 × 109 cells/mL, suitable for assay of complement component activity (H50 units) and whole complement activity (CH50 units), Buffered aqueous cell suspension) E9383
Gelatin veronal buffer G6514
Gelatin veronal buffer-EDTA G9660
back to top
back to Complement Proteins