Plasmin, E.C. 3.4.21.7
Product Name |
Product # |
Plasmin from porcine blood |
P8644 |
|
Specificity:
|
|
Preferential cleavage: Lys-Xaa > Arg-Xaa; higher selectivity than trypsin. Converts fibrin into soluble products
|
|
Synthethic Substrates:
|
|
D-Ala-Leu-Lys-7-amido-4-methylcoumarin, A8171 Boc-Val-Leu-Lys-7-amido-4-methylcoumarin, B1136 Z-Ala-Ala-Lys 4-methoxy-b-naphthylamide formate salt, C2147 Gly-Arg-p-nitroanilide dihydrochl, G8148 D-Ile-Phe-Lys p-nitroanilide, I6886 N-(p-Tosyl)-Gly-Pro-Lys 4-nitro, T6140 D-Val-Leu-Lys 4-nitroanilide dihydrochloride, V0882
|
|
Inhibitors:
|
|
e-Amino-n-caproic acid, A2504 e-Amino-n-caproic acid, A7824 4-(2-Aminoethyl) benzenesulfonyl fluoride hydrochloride, A8456 Antipain, A6191 a2-Antiplasmin, A0914 Antithrombin III, Human, A2221 Antithrombin III, Bovine, A9141 Aprotinin, A1153 Bdellin, B3906 Complement C1 esterase inhibitor, C2412 Complement C1 esterase inhibitor, E0518 3,4-Dichloroisocoumarin, D7910 Diisopropyl fluorophosphate, D0879 Elastatinal, E0881 Gabexate mesylate, G2417 Leupeptin, L2023 a2-Macroglobulin, M6159 Phenylmethylsulfonyl fluoride, P7626 Urinary trypsin inhibitor fragment, U4751
|
back to top back to Proteolytic Enzymes Guide
Prolidase, E.C. 3.4.13.9
Product Name |
Product # |
Prolidase porcine kidney |
P6675 |
|
Specificity:
|
|
Hydrolysis of Xaa Pro dipeptides; also acts on aminoacyl-hydroxyproline analogs. No action on Pro-Pro
|
back to top back to Proteolytic Enzymes Guide
Pronase E
Product Name |
Product # |
Protease Streptomyces griseus, powder Type XIV approx. 4 units/mg solid |
P5147 |
Protease Streptomyces griseus |
P6911 |
Protease Streptomyces griseus, powder approx. 4 units/mg solid Embryo, tested |
P8811 |
|
Specificity:
|
|
Nonspecific Protease mixture
|
|
Inhibitors:
|
|
Diisopropyl fluorophosphate, D0879 EDTA, ED2SS
|
back to top back to Proteolytic Enzymes Guide
Prostate Specific Antigen, E.C. 3.4.21.77
Product Name |
Product # |
Prostate specific antigen from Human Semen, minimum 95% (SDS-PAGE) Buffered aqueous solution |
P3338 |
Prostate specific antigen from Human Semen, minimum 99% (SDS-PAGE) Buffered aqueous solution |
P3235 |
|
Specificity:
|
|
Preferential cleavage: Tyr-Xaa
|
back to top back to Proteolytic Enzymes Guide
Protease, Alkalophilic from Streptomyces griseus
Product Name |
Product # |
Protease Streptomyces griseus |
P0652 |
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Aspergillus (Flavourzyme), E.C. 3.4.11.1
Product Name |
Product # |
Protease from Aspergillus oryzae |
P6110 |
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Aspergillus saitoi (Molsin)
Product Name |
Product # |
Protease from Aspergillus saitoi |
P2143 |
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Aspergillus sojae (Protease IP)
Product Name |
Product # |
Protease from Aspergillus sojae |
P7026 |
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Bacillus licheniformis (alkaline protease)
Product Name |
Product # |
Protease Bacillus licheniformis |
P5459 |
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Bacillus licheniformis (Alcalase)
Product Name |
Product # |
Protease Bacillus licheniformis |
P4860 |
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Bacillus polymyxa
Product Name |
Product # |
Protease from Bacillus polymyxa |
P6141 |
|
Specificity:
|
|
Broad specificity, used for cell dissociation
|
back to top back to Proteolytic Enzymes Guide
Protease from Bacillus sp (Savinase)
Product Name |
Product # |
Protease Bacillus sp. |
P3111 |
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Bacillus sp (Esperase)
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease from Rhizopus sp.
|
Specificity:
|
|
Broad specificity industrial enzyme preparation
|
back to top back to Proteolytic Enzymes Guide
Protease S
Product Name |
Product # |
Protease S Pyrococcus furiosus |
P6361 |
|
Specificity:
|
|
Broad specificity thermostable protease
|
back to top back to Proteolytic Enzymes Guide
Proteasomes
Product Name |
Product # |
Proteasome Fraction II Hela S3 cell extract human |
F3926 |
Proteasome Fraction II from rabbit |
F4051 |
26S Proteasome Fraction from rabbit |
P3863 |
20S Proteasome from rabbit |
P3988 |
|
Specificity:
|
|
Large protease complex directed by ubiqutination
|
|
Synthethic Substrates:
|
|
Z-Leu-Leu-Glu-7-amido-4-methylc, C0483 Z-Leu-Leu-Leu-7-amido-4-methylcoumarin, C0608 N-Succinyl-Leu-Leu-Val-Tyr-7-amido-4-trifluoromethylcoumarin, S4939
|
|
Inhibitors:
|
|
Cbz-Leu-Leu-Phe-al, C0358 Cbz-Leu-Leu-Leu-B(OH)2, C9857 Lactacystin, L6785 clasto-Lactacystin beta-lactone, L7035
|
back to top back to Proteolytic Enzymes Guide
Proteinase from Aspergillus oryzae (Protease M)
Product Name |
Product # |
Proteinase from Aspergillus oryzae |
P4032 |
back to top back to Proteolytic Enzymes Guide
Proteinase 3
Product Name |
Product # |
Proteinase 3 human, minimum 95% (PAGE), PR3 Ag |
P0615 |
back to top back to Proteolytic Enzymes Guide
Proteinase A, E.C. 3.4.23.6
Product Name |
Product # |
Proteinase A from baker's yeast (S. cerevisiae) |
P8892 |
back to top back to Proteolytic Enzymes Guide
Proteinase K, E.C. 3.4.21.64
Product Name |
Product # |
Proteinase K Tritirachium album, Lyophilized powder |
P2308 |
Proteinase K Tritirachium album, Buffered aqueous glycerol solution |
P4850 |
Proteinase K Tritirachium album, Lyophilized powder 1-3 unit/mg solid |
P8044 |
Proteinase K Tritirachium album, Lyophilized powder minimum 30 units/mg protein (biuret) |
P5056 |
Proteinase K Tritirachium album, minimum 500 units/mL Buffered aqueous glycerol solution |
P5568 |
Proteinase K Tritirachium album, Lyophilized powder minimum 30 units/mg protein |
P6556 |
|
Specificity:
|
|
Hydrolysis of keratin, and of other proteins with subtilisin-like specificity. Hydrolyses peptide amides
|
|
Inhibitors:
|
|
Diisopropyl fluorophosphate, D0879 Phenylmethylsulfonyl fluoride, P7626
|
back to top back to Proteolytic Enzymes Guide
Protein C, E.C. 3.4.21.69
Product Name |
Product # |
Protein C human |
P2562 |
Protein C from human plasma |
P2200 |
|
Specificity:
|
|
Cleaves factors Va and VIIIa
|
back to top back to Proteolytic Enzymes Guide
Pyroglutamate aminopeptidase, E.C. 3.4.19.3
Product Name |
Product # |
Pyroglutamate Aminopeptidase Pyrococcus furiosus |
P6236 |
|
Specificity:
|
|
Release of an N-terminal pyroglutamyl group from a polypeptide, the second amino acid generally not being Pro
|
back to top back to Proteolytic Enzymes Guide
Renin, E.C. 3.4.23.15
|
Specificity:
|
|
Cleavage of Leu-bond in angiotensinogen to generate angiotensin I
|
|
Inhibitors:
|
|
Pepstatin A, P5318 D-His-Pro-Phe-His-Leu-psi-(CH2NH)-Leu- Val-Tyr, H6137
|
back to top back to Proteolytic Enzymes Guide
Rennin, E.C. 3.4.23.4
Product Name |
Product # |
Rennin calf stomach |
R4877 |
|
Specificity:
|
|
Broad specificity similar to that of pepsin A. Clots milk by cleavage of a single Ser-Phe105-|-Met-Ala bond in k-chain of casein
|
back to top back to Proteolytic Enzymes Guide
Streptokinase, E.C. 3.4.99.0
Product Name |
Product # |
Streptokinase beta;-hemolytic Streptococcus (Lancefield Group C), Buffered aqueous solution approx. 100,000 units/mg protein |
S8026 |
Streptokinase beta;-hemolytic Streptococcus (Lancefield Group C), Lyophilized powder minimum 3,500 units/mg solid |
S3134 |
back to top back to Proteolytic Enzymes Guide
Subtilisin, E.C. 3.4.21.62
Product Name |
Product # |
Subtilisin A |
P5380 |
|
Specificity:
|
|
Hydrolysis of proteins with broad specificity for peptide bonds, and a preference for a large uncharged residue in P1. Hydrolyses peptide amides
|
back to top back to Proteolytic Enzymes Guide
Thermolysin, E.C. 3.4.24.27
Product Name |
Product # |
Thermolysin Bacillus thermoproteolyticus rokko |
T7902 |
|
Specificity:
|
|
Preferential cleavage: Xaa-Leu > Xaa-Phe
|
|
Inhibitors:
|
|
Diethylenetriaminepentaacetic acid Free Acid, D6518 EDTA, ED2SS EGTA, E4378 a2-Macroglobulin, M6159 1,10-Phenanthroline Monohydrate, P9375 Phosphoramidon, R7385
|
back to top back to Proteolytic Enzymes Guide
Thrombin, E.C. 3.4.21.5
Product Name |
Product # |
Thrombin bovine plasma, Lyophilized powder minimum 700 NIH units/mg protein (Bradford) |
T9681 |
Thrombin from human plasma, suitable (For routine use in thrombine time test) Lyophilized powder |
T9010 |
Thrombin from human plasma, cell culture, tested Lyophilized powder approx. 1,000 NIH units/mg protein (E1%18.3 at 280 nm) |
T4393 |
Thrombin bovine plasma, powder cell culture, tested |
T9549 |
Thrombin from rat plasma, Lyophilized powder minimum 1,000 units/mg protein (biuret) |
T5772 |
Thrombin from murine plasma, Lyophilized powder approx. 1,000 NIH units/mg protein (biuret) |
T8397 |
Thrombin from human plasma, Lyophilized powder 50-300 NIH units/mg protein |
T9677 |
Thrombin from human plasma, Lyophilized powder 50-300 NIH units/mg protein |
T0553 |
Thrombin from human plasma, suitable (For routine use in the thrombin time test) Lyophilized powder |
T8885 |
Thrombin from human plasma, Lyophilized powder |
T1063 |
Thrombin from human plasma, Lyophilized powder minimum 2,000 NIH units/mg protein (E1%18.3 at 280 nm) |
T6884 |
Thrombin from human plasma, suitable (For routine use in the thrombin time test.) Lyophilized powder |
T9135 |
Thrombin from human plasma, Lyophilized powder approx. 1,000 NIH units/mg protein (E1%18.3 at 280 nm) |
T7009 |
Thrombin bovine plasma, Lyophilized powder 600-1,500 NIH units/mg protein (biuret) |
T6634 |
Thrombin bovine plasma, Lyophilized powder 175-350 NIH units/mg protein (biuret) |
T4265 |
Thrombin bovine plasma, minimum 125 NIH units/mg protein (biuret) |
T9000 |
Thrombin bovine plasma, Lyophilized powder 50-175 NIH units/mg protein (biuret) |
T4648 |
Thrombin bovine plasma, Lyophilized powder 50-150 NIH units/mg protein |
T3399 |
Thrombin bovine plasma, Lyophilized powder |
T7513 |
|
Specificity:
|
|
Selective cleavage of Arg-Gly bonds in fibrinogen to form fibrin and release fibrinopeptides A and B
|
|
Synthethic Substrates:
|
|
N-Benzoyl-Phe-Val-Arg 4-methoxy-b-naphthylamide hydrochloride, B1260 Boc-b-benzyl-Asp-Pro-Arg-7-amido-4-methylcoumarin hydrochloride, B4028 N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride, B7632 Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride, B9385 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride monohydrate, M4149 Sar-Pro-Arg p-nitroanilide dihydrochloride, S9009 N-p-Tosyl-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrochloride, T0273 N-(p-Tosyl)-Gly-Pro-Arg p-nitroanilide acetate salt, T1637 Thrombin generation chromogenic substrate, T3068
|
|
Inhibitors:
|
|
N-Acetyl-eglin C, E7888 4-(2-Aminoethyl) benzenesulfonyl fluoride hydrochloride, A8456 Antipain, A6191 a2-Antiplasmin, A0914 Antithrombin III, Bovine, A9141 Antithrombin III, Human, A2221 Antithrombin III, Rat, A6397 3,4-Dichloroisocoumarin, D7910 Diisopropyl fluorophosphate, D0879 Gabexate mesylate, G2417 Hirudin, H0393 Hirudin, H7016 Hirudin, H7380 Hirudin, H9022 Leupeptin, L2023 a2-Macroglobulin, M6159 N a-(2-Naphthalenesulfonylglycyl)-4-amidino-DL-phenylalaninepiperidide, N8514 Phenylmethylsulfonyl fluoride, P7626 N a-p-Tosyl-L-lysine chloromethyl ketone hydrochloride, T7254 D-Val-Leu-Lys-Chloromethyl ketone, V3763
|
back to top back to Proteolytic Enzymes Guide
Tissue Plasminogen Activator
Product Name |
Product # |
Tissue plasminogen activator from human melanoma cells |
T7776 |
|
Specificity:
|
|
Specific cleavage of Arg-Val bond in plasminogen to form plasmin
|
|
Synthethic Substrates:
|
|
Tissue Plasminogen Activator Substrate, T2943
|
back to top back to Proteolytic Enzymes Guide
Trypsin, E.C. 3.4.21.4
Product Name |
Product # |
Trypsin from bovine pancreas, suitable for protein sequencing Lyophilized powder |
T8658 |
Trypsin Gadus morhua (Atlantic cod), Type XX-S 5,000-10,000 units/mg protein Lyophilized powder |
T9906 |
Trypsin from bovine pancreas, Essentially salt-free, lyophilized powder 10,000-15,000 BAEE units/mg protein |
T8802 |
Trypsin from bovine pancreas, Essentially salt-free, lyophilized powder min. 10,000 BAEE units/mg protein |
T1426 |
Trypsin from bovine pancreas, Type XI Lyophilized powder min. 6,000 BAEE units/mg protein |
T1005 |
Trypsin porcine pancreas, Type IX-S Lyophilized powder 13,000-20,000 BAEE units/mg protein |
T0303 |
Trypsin from human pancreas, Salt-free, lyophilized powder |
T6424 |
Trypsin Acetylated from bovine pancreas, synthetic Type V-S approx. 10,000 BAEE units/mg protein (biuret) |
T6763 |
Trypsin from bovine pancreas, suitable for protein sequencing Lyophilized powder |
T8658 |
Trypsin from bovine pancreas, Lyophilized powder min. 7,500 BAEE units/mg solid |
T4665 |
Trypsin porcine pancreas, tablet 1 mg tablet Type II |
T7168 |
Trypsin from bovine pancreas, Type I approx. 10,000 BAEE units/mg protein |
T8003 |
Trypsin porcine pancreas, Type II-S Lyophilized powder 1,000-2,000 BAEE units/mg solid |
T7409 |
Trypsin, Dimethylated porcine, Proteomics Grade |
T6567 |
TrypZean™, recombinant, expressed in Corn Lyophilized powder minimum 3650 units/mg solid (USP) |
T3568 |
|
Specificity:
|
|
Preferential cleavage: Arg-Xaa , Lys-Xaa
|
|
Synthethic Substrates:
|
|
Na-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride, B7260 Na-Benzoyl-DL-arginine-7-amido-4-methylcoumarin hydrochloride, B7385 BAEE, B4500 Na-Benzoyl-L-arginine 4-methoxy-b-naphthylamide hydrochloride, B0885 Na-Benzoyl-DL-arginineb-naphthylamide hydrochloride, B4750 Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride, B3133 Na-Benzoyl-DL-arginine 4-nitroanilide hydrochloride, B4875
|
|
Inhibitors:
|
|
p-Aminobenzamidine dihydrochloride, A7148 4-(2-Aminoethyl) benzenesulfonyl fluoride hydrochloride, A8456 Antipain, A6191 a2-Antiplasmin, A0914 Antithrombin III, Human, A2221 Antithrombin III, Bovine, A9141 a1-Antitrypsin, A6150 a1-Antitrypsin, A9024 Aprotinin, A1153 Bdellin, B3906 Benzamidine Hydrochloride, B6506 Diisopropyl fluorophosphate, D0879 3,4-Dichloroisocoumarin, D7910 Ecotin, E1398 Gabexate mesylate, G2417 Leupeptin, L2023 a2-Macroglobulin, M6159 Phenylmethylsulfonyl fluoride, P7626 N a-p-Tosyl-L-lysine chloromethyl ketone hydrochloride, T7254 Trypsin Inhibitor, T0256 Trypsin Inhibitor, T2011 Trypsin Inhibitor, T4385 Trypsin Inhibitor, T9008 Trypsin Inhibitor, T9128 Trypsin Inhibitor, T9253 Trypsin Inhibitor, T9378 Trypsin-chymotrypsin inhibitor, T9777
|
back to top back to Proteolytic Enzymes Guide
Tryptase, E.C. 3.4.21.59
Product Name |
Product # |
Tryptase from human lung |
T7063 |
|
Specificity:
|
|
Preferential cleavage: Arg-Xaa, Lys-Xaa , but with more restricted specificity than trypsin
|
back to top back to Proteolytic Enzymes Guide
Urokinase, E.C. 3.4.21.73
Product Name |
Product # |
Urokinase from human urine |
U0633 |
|
Specificity:
|
|
Specific cleavage of Arg-Val bond in plasminogen to form plasmin
|
|
Inhibitors:
|
|
Glu-Gly-Arg-chloromethyl ketone, G2917
|
back to top back to Proteolytic Enzymes Guide
|