Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Cloning & Expression
Genotypes of Selected E. coli Strains
Genotypes of Selected E. coli Strains
BL21 E. coli B F ompT gal [E. coli B is naturally dcm and lon] hsdSB
BL21(DE3) E. coli B F ompT gal [E. coli B is naturally dcm and lon] hsdSB with DE3, a λ prophage carrying the T7 RNA polymerase gene and lacIQ
C600 F [e14– (McrF) or e14+ (McrF+)] thr-1 leuB6 thi-1 lacY1 glnV44 rfbD1 fhuA21
CJ236 F Δ(HindIII)::cat (Tra+ Pil+ CamR )/ ung-1 relA1 dut-1 thi-1 spoT1 mcrA
GC5 F´ endA1 hsdR17 (rKmK+) glnV44 thi-1 recA1 gyrA (Nalr) relA1 Δ(lacIZYA-argF)U169 (φ80dlacΔ(lacZ)M15 fhuA
GM48 F thr leu thi lacY galK galT ara fhuA tsx dam dcm glnV44
HB101 F Δ(gpt-proA)62 leuB6 glnV44 ara-14 galK2 lacY1 Δ(mcrC-mrr) rpsL20 (Strr) xyl-5 mtl-1 recA13
JM83 F ara Δ(lac-proAB) rpsL (Strr)[φ80 dlacΔ(lacZ)M15] thi
JM101 F´traD36 proA+B+ lacIq Δ(lacZ)M15/ Δ(lac-proAB) glnV thi
JM103 F´ traD36 lacIqΔ(lacZ)M15 proA+B+/endA1 glnV sbcBC thi-1 rpsL (Strr) Δ(lac-pro) (P1) (rK–mK+ rP1+ mP1+)
JM105 F´ traD36 lacIqΔ(lacZ)M15 proA+B+/thi rpsL (Strr) endA sbcB15 sbcC hsdR4 (rK–mK+) Δ(lac-proAB)
JM107 F´ traD36 lacIq Δ(lacZ)M15 proA+B+/e14(McrA) Δ(lac-proAB) thi gyrA96 (Nalr) endA1 hsdR17 (rK mK+) relA1 glnV44
JM109 F´traD36 proA+B+ lacIq Δ(lacZ)M15/ Δ(lac-proAB) glnV44 e14- gyrA96 recA1 relA1 endA1 thi hsdR17
JM110 F´ traD36 lacIqΔ(lacZ)M15 proA+B+IrpsL (Strr) thr leu thi lacY galK galT ara fhuA dam dcm glnV44 Δ(lac-proAB)
K802 F e14- (McrA-) lacY1 or Δ(lac)6 glnV44 galK2 galT22 rfbD1 metB1 mcrB1 hsdR2 (rKmK+)
LE392 F e14 (McrA–) hsdR514 (rKmK+) glnV44 supF58 lacY1 or Δ(lacIZY)6 galK2 galT22 metB1 trpR55
MC1061 F araD139 Δ(ara-leu)7696 galE15 galK16 Δ(lac)X74 rpsL (Strr) hsdR2 (rKmK+) mcrA mcrB1
MM294 F endA1 hsdR17 (rKmK+) glnV44 thi-1 relA1 rfbD1 spoT1
NM477 C600 Δ(hsdMS-mcrB)5 (rKmK+ McrBC)
NM522 F´proA+B+ lacIq Δ(lacZ)M15/ Δ(lac-proAB) glnV thi-1 Δ(hsdS-mcrB)5
NM554 MC1061 recA13
NM621 F hsdR (rKmK+) mcrA mcrB glnV44 recD1009
RR1 HB101 RecA+
χ1776 F fhuA53 dapD8 minA1 glnV44 Δ(gal-uvrB)40 minB2 rfb-2 gyrA25 (Nalr) thyA142 oms-2 metC65 oms-1 (tte-1) Δ(bioH-asd)29 cycB2 cycA1 hsdR2 (rK mK+) mcrB1
References
  1. New England Biolabs. E. coli Strain Genotypes. http://www.neb.com/nebecomm/tech_reference/restriction_enzymes/ecoli_genotypes.asp (Oct. 31, 2005)
  2. Wertman, K.F. et al. (1986) Gene 49, 253–262.
  3. Yanisch–Perron, C., Viera, J. and Messing, J. (1985) Gene 33, 103–119.
  4. Sambrook, J., Fritsch, E.F. and Maniatis, T. (1989). Molecular Cloning: A Laboratory Manual, (2nd ed.). Cold Spring Harbor: Cold Spring Harbor Laboratory Press.
  5. Huynh,T.V. et al. (1985). In D.M. Glover (Ed.), DNA Cloning Vol. 1, (pp. 56–110). Oxford, England: IRL Press Limited.
  6. Raleigh, E.A. et al. (1988) Nucl. Acids Res. 16, 1563–1575.
  7. Woodcock, D.M. et al. (1989) Nucl. Acids Res. 17, 3469–3478.
  8. Raleigh, E.A., Lech, K. and Brent, R. (1989). In F.M. Ausebel et al. (Eds.), Current Protocols in Molecular Biology (p. 1.4). New York: Publishing Associates and Wiley Interscience.
  9. Berlyn, M.K.B. (1996). In F.C. Niedhardt et al. (Ed.), Escherichia coli and Salmonella: cellular and molecular biology, (2nd ed.), Vol. 2, (pp. 1715–1902). ASM Press.
  10. Miller, J.H. (1972). Experiments in Molecular Genetics. Cold Spring Harbor: Cold Spring Harbor Laboratory Press.
  11. Whittaker, P.A. et al. (1989) Nucl. Acids Res. 16, 6725–6736.
  12. Murray, N.E. et al. (1977) Mol. Gen. Genet. 150, 53–61.
  13. Palmer, B.R. and Marinus, M.G. (1994) Gene 143, 1–12.
  14. Boyer, H.W, and Roulland–Dussoix, D. (1969) J. Mol. Biol. 41, 459.
  15. Silhavy, T.J. et al. (1984) Experiments with Gene Fusions (pp. xi–xii) Cold Spring Harbor: Cold Spring Harbor Laboratory.
  16. Bullock, W.O. et al. (1987) BioTechniques 5, 376–378.
  17. Maurizi, M.R. et al. (1985) J. Bacteriol. 164, 1124–1135.
  18. Studier, F.W. et al. (1990). In D.V. Goeddel (Ed.), Methods in Enzymology Vol. 185, (pp. 60–89). San Diego: Academic Press.
  19. Kelleher, J. and Raleigh, E.A. (1991) J. Bacteriol. 173, 5220–5223.
  20. Woodcock, D.M. et al. (1989) Nucl. Acids Res. 17, 3469–3478.
  21. Palmer, B.R. and Marinus, M.G. (1994) Gene 143, 1–12.
  22. Yanisch-Perron, C., Viera, J. and Messing, J. (1985) Gene 33, 103–119.
  23. Messing, J. (1979) Recombinant DNA Technical Bulletin (NIH) 2, 43–48.
  24. Gough, J. and Murray, N. (1983) J. Mol. Biol. 166, 1–19.
  25. Baker, T.A. et al. (1984) Proc. Nat. Acad. Sci. USA 81, 6779–6783.
  26. Grossman, A.D. et al. (1983) Cell 32, 151–159
  27. Chung, C.H. and Goldberg, A.L. (1981) Proc. Nat. Acad. Sci. USA 78, 4931–4935.
  28. Straus et al. (1988) Genes Dev. 2, 1851–1858.
  29. Kowit, J.D. and Goldberg, A.L. (1977) J. Biol. Chem. 252, 8350–8357.
  30. Silber, K.R. and Sauer R.T. (1994) Mol. Gen. Genet. 242, 237–240.
  31. Elish et al. (1988) J. Gen. Microbiol. 134, 1355–1364.
  32. Kunkel, T.A. et al. (1987). In R. Wu and L. Grossman (Eds.), Methods in Enzymology Vol. 154, (pp. 367–382). San Diego: Academic Press.
Back to top
Back to Cloning and Expression