Electropherogram revealing >600 bases of sequence from pCMV-SPORT-ß-gal purified from GenElute HP Plasmid Midiprep Kit. Cycle Sequencing was performed using 500 ng template, a T7 sequencing primer and ABI BigDye™ terminator chemistry. Sequencing reactions were resolved on an ABI Prism XL instrument with a 48 cm gel cassette containing 4.5% AutoPAGE Plus acrylamide at 2.4 kV, 48°C for 7 hours.