DNA & RNA Purification
Plasmid Purification
Our GenElute™ plasmid kits provide reliable isolation of high quality plasmid DNA. These kits are:
  • Designed with advanced silica bind and elute technology
  • Compatible with both vacuum or spin format
  • Formulated without phenol/chloroform
These products allow researchers to maximize the reproducibility of their workflow and minimize their time spent on obtaining plasmid DNA for transfection, sequencing, PCR and other downstream applications.
Product No. Product Description
Starting Amount
Time Required
Elution Volume
Expected Yield
Miniprep
PFM10 (10 preps)
PFM50 (50 preps)
PFM250 (250 preps)
GenElute™ Five-Minute Plasmid Miniprep Kit
400 µL
5 min
40 µL
Up to 5 µg
PLN10 (10 preps)
PLN70 (70 preps)
PLN350 (350 preps)
GenElute™ Plasmid Miniprep Kit
1.5mL
<30 min
100 µL
Up to 20 µg
NA0150S (10 preps)
NA0150 (70 preps)
NA0160 (350 preps)
GenElute™ HP Plasmid Miniprep Kit
1-5 mL
<30 min
100 µL
Up to 25 µg
NA9604 (4 x 96 wells)
GenElute™ HP 96 Well Plasmid Miniprep Kit
1.3 mL/well
45 min
100 µL
Up to 10 µg/well
Midiprep
NA0200S (4 preps)
NA0200 (25 preps)
GenElute™ HP Plasmid Midiprep Kit
50 mL
30 min
1 mL
Up to 350 µg
PLED35 GenElute™ Endotoxin-Free Plasmid Midiprep Kit
5-40 mL
<95 min
1 mL
Up to 250 µg
Maxiprep
NA0300S (4 preps)
NA0300 (10 preps)
NA0310 (25 preps)
GenElute™ HP Plasmid Maxiprep Kit
150 mL
30 min 3-5 mL
Up to 1.2 mg
NA0400S (4 preps)
NA0400 (10 preps)
NA0410 (25 preps)
GenElute™ HP Endotoxin-Free Plasmid Maxiprep Kit 150 mL 40 min 3-5 mL
Up to 1.2 mg
Megaprep
NA0600 (5 preps)
GenElute™ HP Endotoxin-Free Plasmid Megaprep Kit
200 mL–1 L
90 min
20 mL
Up to 5 mg
Gigaprep
NA0800 (5 preps)
GenElute™ HP Endotoxin-Free Plasmid Gigaprep Kit
Up to 2.5 L
2 hrs
50 mL
Up to 15 mg