DNA & RNA Purification
Gel Purification
We provide PCR clean-up, agarose spin columns and minus EtBr spin columns for purification of single or double stranded PCR amplification fragments; as well as extraction of DNA fragments from Ethidium Bromide (EtBr) stained agarose gels.
Applications for these kits would include enzymatic reactions, automated sequencing, cloning, microarray analysis, ligation, restriction enzyme digestions, and PCR.
Cat. # Product Description Starting Amount Time Required Expected Yield
PCR Clean-up
11732676001 (250 preps)
11732668001 (50 preps)
High Pure PCR Product Purification Kit 100 μl <10 minutes for PCR product, and purify DNA from agarose gel slices in <20 minutes  
4983912001 (200 preps)
4983955001 (50 preps)
High Pure PCR Cleanup Micro Kit Varies 10 min 85% recovers up to 20μg DNA
NA1020 (70 preps) GenElute™
PCR Clean-Up Kit
100 µL or 10 µg of DNA <10 min Up to 95% recovery
PCR9601 (1 x 96-well)
PCR9604 (4 x 96-well)
GenElute™ 96Well PCR Clean-up Kit <100 µL PCR reaction 45 min per plate 75 – 95% recovery
D1947
(8RXN / 48RXN / 96RXN)
Diffinity RapidTip® for PCR Purification 25 µL 1 min Up to 95% recovery
D1947L
(8RXN / 48RXN / 96RXN)
Diffinity RapidTip® for PCR Purification, for Rainin LTS® 25 µL 1 min Up to 95% recovery
D2947
(8RXN / 48RXN / 96RXN)
Diffinity RapidTip®2 with polymerase removal 50 µL 1 min Up to 95% recovery
D2947L
(8RXN / 48RXN / 96RXN)
Diffinity RapidTip®2 with polymerase removal, for Rainin LTS® 50 µL 1 min Up to 95% recovery
Gel Extraction
11696505001 Agarose Gel DNA Extraction Kit 100 mg gel slice 60 min Varies depending on size range of recovered DNA
NA1111 (70 preps) GenElute™ Gel Extraction Kit Up to 3.5 g gel slice N/A Up to 80% recovery
56500 (70 preps) GenElute™ Agarose Spin Columns <200 mg gel slice 10 min Up to 80% recovery
56501 (70 preps) GenElute™ Minus EtBr Spin Columns <200 mg gel slice 10 min Up to 70% recovery