DNA & RNA Purification
RNA Purification
Our selection of RNA purification kits offers simple solutions for RNA isolation. The selection of RNA purification kits:
  • Utilize advanced silica bind and elute technology
  • Are formulated without phenol or chloroform
  • Include kits designed specifically for FFPE, plasma/serum, single cell, plant, water and other sample types are available.
  • Include kits for RNA/DNA co-purification
The GenElute™ selection also includes stand-alone reagents for RNA isolation and RNA/DNA co-purification.
Product No. Product Description Starting Amount Time Required Expected Yield
Total RNA
12033674001 High Pure RNA Tissue Kit Varies depending on method used to prepare tissue 30 min Varies depending on tissue type
RTN10 (10 preps)
RTN70 (70 preps)
RTN350 (350 preps)
GenElute™ Mammalian Total RNA Miniprep Kit Up to 107 mammalian cells or 40 mg of tissue 30 min Up to 150 µg of total RNA
RNB100-50RXN
RNB100-100RXN
GenElute™ Universal Total RNA Purification Kit Up to 106 mammalian cells or 25 mg of tissue 20 min Up to 50 µg of total RNA
RNB200-8RXN GenElute™ Total RNA Purification Maxi Kit Up to 5x107 mammalian cells or 1 g of tissue 40 min Up to 1.5 mg
RNB300-50RXN GenElute™ Single Cell RNA Purification Kit Single cell to 2x10cells 20 min Varies depending on starting amount
RTN9602 (96-well x 2)
RTN9604 (96-well x 4)
GenElute™ 96 Well Total RNA Purification Kit Up to 107 cultured cells or 30 mg of tissue 70 min 50-200 ng/µL
mRNA
11934333001 mRNA Isolation Kit for Blood/Bone Marrow 1.5-5 mL blood/bone marrow aspirate 90 min 50-200 ng/mL blood
11741985001 mRNA Isolation Kit Varies depending on sample type 30 min Varies depending on sample type
11787896001 mRNA Capture Kit Varies depending on sample type 15 min Varies depending on sample type
MRN10 (10 preps)
MRN70 (70 preps)
GenElute™ mRNA Miniprep Kit 5-500 µg total RNA 40 min 1-5% of starting total RNA
DMN10 (10 preps)
DMN70 (70 preps)
GenElute™ Direct mRNA Miniprep Kit Up to 107 mammalian cells or 50 mg of tissue 60 min Up to 5 µg
microRNA
05080576001 High Pure miRNA Isolation Kit Depends on sample type 30 min Varies depending on tissue type
SNC10 (10 preps)
SNC50 (50 preps)
mirPremier® microRNA Isolation Kit Up to 107 cultured cells or 40 mg of tissue 30 min Up to 20 µg
R4533-50ML
R4533-100ML
R4533-200ML
RNAzol® RT 1 mL sufficient for 107 cells or 100 mg of tissue < 1 hour 0.1 µg/ µL of microRNA  
RNA/DNA Co-Purification
11934317001 RNA/DNA Stabilization Reagent for Blood/Bone Marrow 16.5 ml to 55 ml of lysate    
RDP300-50RXN GenElute™ RNA/DNA/Protein Plus Purification Kit Up to 5 x 106 cells
or 25 mg of tissue
30 min For 106 HeLa cells, avg yield of 15 μg RNA, 8 μg DNA, and 150 μg protein
T9424-25ML
T9424-100ML
T9424-200ML
TRI Reagent® Up to 107 cells or 100 mg of tissue 90 min 5-15 µg/106 cells; 1-10 µg/mg tissue
E4788 (50 preps) Plant RNA/DNA Purification Kit Up to 5 x 106 cells  or 100 mg of tissue 30 min Up to 50 µg for RNA; 15 µg for gDNA
RDP100-50RXN GenElute™ Water RNA/DNA Purification Kit – 0.45µm 10 – 100 mL 45 min 100 µL
RDP200-50RXN GenElute™ FFPE RNA/DNA Purification Plus Kit Up to 10 mg block or 4 sections <20 µM thick 1 – 4 hrs; varies depending on incubation time and formalin crosslink-reversal Up to 35 µg for RNA; 10 µg for DNA
Special Samples
03270289001 High Pure RNA Paraffin Kit Varies depending on sample type 2 hrs without over night incubation time Varies depending on sample type
04823125001 High Pure FFPE RNA Micro Kit 1-10 μm FFPE sections, colon, breast, liver, kidney, spleen of mammalian species 60 min without 3 hour incubation 1.5 - 3.5 μg/5 μm section
STRN10 (10 preps)
STRN50 (50 preps)
STRN250 (250 preps)
Spectrum™ Plant Total RNA Kit Up to 100 mg of ground plant tissue 30 min 20-60 µg
E4913 (50 preps) Plant/Fungi RNA Purification Kit Up to 50 mg plant tissue; 1 x 106 cells; 50 mg of fungi 30 min Up to 60 µg; varies depending on starting material
RNB400-50RXN GenElute™ FFPE RNA Purification Kit Up to 25 mg block or 5 sections of 20 µm thick paraffin slices 1 – 4 hrs; varies depending on incubation time and formalin crosslink-reversal Up to 50 µg
RNB500-50RXN GenElute™ Plasma/Serum RNA Purification Kit 50 – 200 µL 15 – 20 min Varies depending on specimen
RNB600-20RXN GenElute™ Plasma/Serum RNA Purification Midi Kit 250 µL – 1.5 mL 35 – 40 min Varies depending on specimen
RNB700-10RXN GenElute™ Plasma/Serum RNA Purification Maxi Kit 2 mL – 5 mL 35 – 40 min Varies depending on specimen