Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MystiCq® microRNA RT-qPCR Compatible Instrument Table
MystiCq microRNA SYBR Green qPCR ReadyMixes are ready-to-use mixes containing reference dye optimized for a variety of thermocyclers. Use the chart below to help select the appropriate qPCR ReadyMix for your lab.
Different real-time PCR systems employ different strategies for normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument. MystiCq® microRNA SYBR Green qPCR ReadyMixes contain ROX passive reference dye at concentrations recommended by these (and other) popular cyclers.
Real-time PCR Instrument Name MystiCq microRNA SYBR® Green qPCR ReadyMix™ MystiCq microRNA SYBR Green qPCR ReadyMix Low ROX™ MystiCq microRNA SYBR Green qPCR ReadyMix with ROX
  MIRRM00 MIRRM01 MIRRM02
Applied Biosystems 5700    
Applied Biosystems 7000    
Applied Biosystems 7300    
Applied Biosystems 7500    
Applied Biosystems 7500 Fast    
Applied Biosystems 7700    
Applied Biosystems 7900    
Applied Biosystems 7900 HT    
Applied Biosystems 7900 HT Fast    
Applied Biosystems StepOnePlus™    
Applied Biosystems StepOne™    
Applied Biosystems ViiA™ 7    
Bio-Rad CFX384™    
Bio-Rad® CFX96™    
Bio-Rad MiniOpticon™    
Bio-Rad/MJ Chromo4™    
Bio-Rad/MJ Opticon 2™    
Bio-Rad/MJ Opticon™    
Cepheid SmartCycler®    
Eppendorf Mastercycler® ep realplex    
Eppendorf Mastercycler® ep realplex2 S    
Illumina Eco™ qPCR    
Qiagen/Corbett Rotor-Gene® 3000    
Qiagen/Corbett Rotor-Gene® 6000    
Qiagen/Corbett Rotor-Gene® Q    
Roche LightCycler™ 480    
Stratagene Mx3000P™    
Stratagene Mx3005P™    
Stratagene Mx4000™