Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Plant Molecular Biology
Maize SSR Primer Plate 4
Maize SSR Home
Plate 1
Plate 2
Plate 3
Plate 4 (Searchable Format - 28 Kb PDF)
   Row A   Row B   Row C   Row D   Row E   Row F   Row G   Row H
Plate 4 — Click on Gene Names to view maizegdb.org
Row A back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1154 4/A1 7 558.55
p-bnlg469 4/A2 7 572.56
p-umc2333 4/A3 7 593.4
p-phi116 4/A4 7 611.5
p-umc1786 4/A5 8 10.58
p-umc2042 4/A6 8 30.3
p-umc1414 4/A7 8 48
p-umc1075 4/A8 8 59.88
p-umc1483 4/A9 8 83.1
p-bnlg1194 4/A10 8 105.4
p-umc1304 4/A11 8 128.6
p-umc1974 4/A12 8 153.3
Row B back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1913 4/B1 8 160.8
p-umc1530 4/B2 8 179.5
p-umc2146 4/B3 8 198.4
p-umc2354 4/B4 8 216.2
p-umc1984 4/B5 8 234.8
p-umc1802 4/B6 8 257.67
p-phi100175 4/B7 8 274.9
p-umc2154 4/B8 8 295.3
p-bnlg2046 4/B9 8 315.2
p-umc1343 4/B10 8 337.2
p-umc1846 4/B11 8 357.9
p-umc1340 4/B12 8 377.7
Row C back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1824 4/C1 8 397.86
p-bnlg1152 4/C2 8 414.1
p-umc1728 4/C3 8 432.4
p-bnlg1031 4/C4 8 455.1
p-umc1607 4/C5 8 466.5
p-bnlg1823 4/C6 8 483.4
p-bnlg1828 4/C7 8 506.8
p-umc1005 4/C8 8 526.6
p-umc1673 4/C9 8 546.9
p-umc1069 4/C10 8 571.5
p-umc1663 4/C11 8 608.1
p-bnlg1131 4/C12 8 628.2
Row D back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1957 4/D1 9 0
p-umc1370 4/D2 9 17.7
p-umc1867 4/D3 9 24.3
p-bnlg1288 4/D4 9 43.23
p-bnlg1583 4/D5 9 62.3
p-phi033 4/D6 9 80.3
p-umc1170 4/D7 9 101.1
p-umc2336 4/D8 9 125.7
p-bnlg244 4/D9 9 142.6
p-bnlg1372 4/D10 9 155.16
p-umc1698 4/D11 9 170.4
p-phi022 4/D12 9 191.7
Row E back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1586 4/E1 9 199.7
p-umc2337 4/E2 9 220.1
p-umc2340 4/E3 9 238.4
p-umc1267 4/E4 9 257.6
p-bnlg1209 4/E5 9 273.2
p-umc1107 4/E6 9 285.8
p-bnlg1012 4/E7 9 298
p-umc2121 4/E8 9 315.7
p-umc1231 4/E9 9 342
p-bnlg1091 4/E10 9 351.23
p-umc1494 4/E11 9 373.66
p-umc2343 4/E12 9 385.3
Row F back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc2134 4/F1 9 404.3
p-umc1794 4/F2 9 420.18
p-umc2346 4/F3 9 461.6
p-umc1366 4/F4 9 489.9
p-umc1310 4/F5 9 517.73
p-phi448880 4/F6 9 536.8
p-bnlg619 4/F7 9 554.4
p-umc2347 4/F8 9 578.6
p-umc1137 4/F9 9 603.5
p-umc1982 4/F10 9 633.2
p-umc1380 4/F11 10 16.6
p-phi041 4/F12 10 30.9
Row G back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1291 4/G1 10 58.76
p-umc2053 4/G2 10 76.2
p-umc1576 4/G3 10 97.9
p-umc2034 4/G4 10 120.1
p-umc1337 4/G5 10 143.3
p-bnlg1547 4/G6 10 161.85
p-umc1962 4/G7 10 180.7
p-umc1345 4/G8 10 199.5
p-bnlg1712 4/G9 10 217.8
p-umc2348 4/G10 10 244.6
p-MZETC34 4/G11 10 268.1
p-umc1330 4/G12 10 287.9
Row H back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1154 4/H1 10 308.7
p-bnlg1074 4/H2 10 332.1
p-umc1506 4/H3 10 344.8
p-umc1477 4/H4 10 366.3
p-umc2122 4/H5 10 392.5
p-bnlg2190 4/H6 10 412.3
p-umc1196 4/H7 10 444.8
p-bnlg1839 4/H8 10 466.4
p-umc1877 4/H9 10 488.63
p-umc2021 4/H10 10 505.5
p-umc1038 4/H11 10 525.11
p-umc1645 4/H12 10 578.09
back to top