Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Plant Profiler
St. John's wort (Hypericum perforatum)
Synonyms / Common Names / Related Terms
Amber touch-and-heal, balm-of-warrior's wound, balsana, bassant, Blutkraut, bossant, Calmigen®, corancillo, dendlu, devil's scorge, Eisenblut, flor de São João, fuga daemonum, goatweed hartheu, heofarigo on, herba de millepertius, herba hyperici, Herrgottsblut, Hexenkraut, hierba de San Juan, hipericão, hipérico, hipericon, HP, isorhamnetin, Jarsin, Johanniskraut, klammath weed, Liebeskraut, LI 160, lord God's wonder plant, millepertius pelicao, perforate, pinillo de oro, PM235, pseudohypericin, rosin rose, SJW, SJW extract LI 160, St. John's wort WS 5572, STW 3-VI, tenturotou, Teufelsflucht, touch and heal, Walpurgiskraut (German), witcher's herb, WS 5572.
Mechanism of Action
Pharmacology:
  • Overview: St. John's wort cultivated for medicinal use is harvested upon flowering and is prepared from the aerial portions of the plant. Herbal preparations are then extracted with ethanol or methanol. Notably, the hyperforin, hypericin, and flavonol content may vary widely according to harvest time, plant quality, and region of origin.39 The apparent broad mechanism of St. John's wort is not fully understood, yet biologically active constituents may include hyperforin and adhyperforin (phloroglucinols), hypericin and pseudohypericin (naphthodianthrones), flavonoids, xanthones, oligomeric procyanidines, and amino acids.40,41 Methods for evaluation and standardization of the components are still under active investigation.42,43 St. John's wort's antidepressant activity may be mediated by the serotonergic (5-HT), noradrenergic (NE), and dopaminergic (DA) systems44,34,45,46,47,48, as well as via the neurotransmitters glutamate and γ-aminobutyric acid (GABA)14,35,49,50. Weak activity in vitro suggests a combination of multiple mechanisms51, several of which are presented in the following paragraphs.
  • Antimicrobial effects: MeOH, petroleum ether, CHCl3, and EtOAc extracts of the aerial parts of St. John's wort were assayed for antimicrobial activity. Growth inhibition was observed for the Gram-positive bacteria B. subtilis and B. cereus. The Hypericum extract obtained with EtOAc was the most active. The main constituents of this extract, as determined by HPLC analysis, were flavonoids, hypericins, and hyperforins. Incubation of the selected microorganisms with the pure chemicals resulted in a significant inhibition of their growth by hypericin, hyperforin, and its stable dicyclohexilammonium salt. Flavonoids appeared inactive.1
  • Antioxidant effects: In rats, administration of 1.4mg/kg of scopolamine impaired retrieval memory. This amnesia was associated with elevated MDA and reduced GSH brain levels. In naive rats, which had not been exposed to conditioned fear, scopolamine administration also increased MDA and reduced GSH levels, although with an increase in brain GSHPx activity. Pretreatment of the animals with Hypericum extract (4, 8, and 12mg/kg) resulted in an antioxidant effect through altering brain MDA, GSHPx, and/or GSH level or activity. Low concentrations of Hypericum extract displayed an inverse dose-related relationship of superoxide inhibition.1 Free radical-scavenging activity of Hypericum has been found to correlate with its flavonoid constituents, including hyperoside and quercetin.2,3
  • Anti-proliferative/anti-inflammatory effects: St. John's wort was found to inhibit the growth of, and induce apoptosis in, experimental leukemia and glioma cell lines.52,4 Similar apoptoic activity was observed in mouse tumor models.53 An immunosuppressive effect of hypericin was found in vitro, mediated by inhibition of arachidonic acid and interleukin-6 release, leukotriene B4, and interleukin-1α production.54,55 An immunosuppressive effect, including an inhibition of T-cell proliferation, was observed in vitro and in vivo in response to St. John's wort (in vivo, the effect of Hypericum ointment was compared with the immunosuppressive effect of solar-simulated radiation), which may provide a rationale for the treatment of inflammatory skin disorders with St. John's wort extracts.21 In vitro inhibition of free radical production has also been demonstrated in cell-free and human vascular tissue.3
  • Hyperforin can stimulate IL-8 expression in human intestinal epithelia cells (IEC) and primary hepatocytes. Hyperforin is also able to induce the expression of mRNA for ICAM-1. Hyperforin was shown to induce IL-8 mRNA through an SXR-independent transcriptional activation pathway. IL-8 induction by hyperforin required the activation of AP-1 but not the NF-kappaB transcription factor. Further study revealed that extracellular signal-regulated kinase 1 and 2 (ERK1/2) were required for the hyperforin-induced expression of IL-8.56 Extracts of St. John's wort strongly down-regulate mitogen-mediated tryptophan degradation in a dose-dependent manner. This effect seems to be based on a suppressive activity of H. perforatum on activated immunocompetent cells, resulting in a diminished production of IFN-gamma.57
  • The stable dicyclohexylammonium salt of hyperforin was shown to trigger an apoptosis-associated cytotoxic effect in murine and human tumor cells. Untransformed endothelial cells were only marginally affected. An inhibition of various proteinases instrumental to extracellular matrix degradation was also noted (leukocyte elastase, cathepsin G, and urokinase-type plasminogen activator). In mice that received intravenous injections of C-26 or B16-LU8 cells daily, intraperitoneal administration of hyperforin reduced inflammatory infiltration, neovascularization, lung weight, and size and number of experimental metastases.58
  • Hyperforin has been shown to target components within the G protein signaling cascades involved in the regulation of Ca2+ homeostasis, resulting in regulation of ROS generation and the release of leukocyte elastase from human isolated polymorphonuclear leukocytes.59 Hyperforin inhibited IL-6 release from human astrocytoma cells (U373MG) in vitro. In whole blood, however, hyperforin levels needed to inhibit IL-6 release are about one order of magnitude higher than the hyperforin levels measured in the plasma of rats or humans treated with pharmacologically active doses of St. John's wort or hyperforin.60
  • In vivo, hypericin treatment inhibited NF-kappaB caused accumulation of phosphorylated IkappaBalpha, decreased p50 protein levels, and induced cleavage of p65 protein in U373 and MCF-7 cells.61
  • Antiviral effects: A variety of in vitro studies have documented St. John's wort's antiviral properties.22,23,24,25,26,27,28,29,30,31 A photodynamic mechanism has been proposed for the mechanism of action of HIV inactivation.30,28,62
  • Drug metabolism: St. John's wort and hyperforin were observed to increase mRNAs for the drug metabolizing enzymes CYP3A4, CYP1A1, and CYP1A2, the flavin-containing monooxygenase FMO5, and of the multidrug resistance protein MRP2. CYP4F2 and the reduced nicotinamide adenine dinucleotide dehydrogenase NQO1 were down-regulated. Expression of genes mediating cholesterol biosynthesis was decreased, while facilitated glucose transporters and glycolysis genes were induced, indicating increased glucose metabolism. Changes of a considerable number of additional transcripts corresponded to reports on gene regulation by hypoxia. Endoplasmic reticulum stress-regulated genes involved in unfolded protein response and in protection of cells from apoptosis were down-regulated. Other calcium binding proteins were affected by both treatments, suggesting an increase in intracellular calcium.11
  • Electroencephalogram (EEG) findings: Electrophysiological studies have found enhanced striatal alpha-1 activity early after oral administration, which may indicate an interaction with serotonin reuptake; later increases in delta and beta-2 activity may be correlated with GABA binding and NMDA agonism.63 Enhanced activation in beta-2 regions had been previously reported.64 In a human study, increases in alpha-1, delta, and theta activity was found to increase with St. John's wort.65 It has been suggested that evaluations of EEG activity may provide more insight into the neurophysiology of St. John's wort than examinations of REM sleep.66 However, Sharpley reported that Hypericum increased latency to REM sleep similar to standard antidepressants.67,68
  • Monoamine oxidase inhibitor (MAOI) activity: Early work reported in vitro inhibition of monoamine oxidase (MAO) A and B by hypericin16, as well as other components, such as xanthon and flavonols19. Thiede and Walper found both MAO and catechol-o-methyltransferase (COMT) inhibition.20 Yet these authors, along with others17, concluded that the concentrations causing inhibition were not adequate to explain antidepressant activity. Cott reported hypericin lacked significant MAO inhibition at concentrations to 10mcM14, and based upon other findings, such as a pharmacokinetics study by Staffeldt et al.38, suggested that this inhibition may not be pharmacologically relevant.
  • Serotonin (5-HT), norepinephrine (NE), dopamine (DA) activities: It has been proposed that the activity of St. John's wort is occurs via inhibition of serotonin, norepinephrine, and dopamine synaptic reuptake.34,35,36,37 Müller reported that hyperforin is likely the active component; hyperforin approximated the molar efficacy of standard tricyclic antidepressants, with uniquely similar potency in serotonin, norepinephrine, and dopamine systems.34 Although not definitive, efficacy in behavioral paradigms of depression (learned helplessness, behavioral despair) has correlated with hyperforin content.69 Bhattacharya reported dose-dependent potentiation of serotonin-mediated animal behaviors, with greater effect in the hyperforin-enriched (38.8% hyperforin) CO2 extract. In contrast, the ethanolic extract (4.5% hyperforin) potentiated dopamine-mediated behaviors.45 In addition, significant down-regulation of B-receptor density and an increase in 5-HT2 receptors has been demonstrated in animal cortex following treatment with Hypericum extract.34 The number of both 5-HT1 A and 5-HT2 A receptors was significantly increased, by 50%, compared to controls in another report.70 In the Müller study, the effect on serotonergic receptors varied according to the extract; a methanolic extract led to a significant increase in receptor density, compared to a (nonsignificant) decrease in receptor density found with a hyperforin-enriched CO2 extract.34 A neuroendocrine study in healthy adults demonstrated an increase in cortisol with 600mg oral WS5570 extract, suggesting central norepinephrine or serotonin neurotransmitter activity. The authors posit that hyperforin plasma concentrations vary according to dose of extract and should be considered in evaluating biochemical and clinical activity.71
  • Other neurotransmitter biochemical effects: The mechanism of St. John's wort activity may be different from standard antidepressants.72 Monoamine inhibition may be noncompetitive, via sodium channels, such as enhancement of intracellular Na+ concentrations.18,49,50 Jensen has also suggested that a direct effect on known transporter sites may not be the mechanism.73 In this study, neither hyperforin nor adhyperforin inhibited binding of a cocaine analogue to the dopamine transporter. Benzodiazepine, adenosine, inositol triphosphate, GABA, N-methyl-D-aspartic acid14,15, and cholinergic receptor activity74 may also contribute to psychotropic effects. Likewise, St. John's wort has been found to affect nighttime melatonin levels, modulate cytokine expression55, increase cortisol71, inhibit sigma opioid receptor activity74 and antagonize naloxone44.
  • After seven days of treatment with St. John's wort, an increase was noted in the dopamine metabolite plasma dihydroxyphenylacetic acid.75 Wong et al. suggest that antidepressant treatment by St. John's wort may be due to changes in gene expression in a discrete brain region, based on the identification of common differentially regulated genes between St. John's wort and a conventional antidepressant (imipramine) in the rat hypothalamus.76
Pharmacodynamics/Kinetics:
  • Drug concentration levels: After oral administration of hyperforin 300mg, steady state plasma concentrations were approximately 100ng/mL.13
  • After long-term dosing of Hypericum extract (300mg three times daily), steady state was reached after four days.38
  • Time to peak: After single oral doses of 300, 900, and 1,800mg of Hypericum extract, the median maximal plasma levels were 1.5, 4.1, and 14.2ng/mL, respectively, for hypericin and 2.7, 11.7, and 30.6ng/mL, respectively, for pseudohypericin.38
  • In humans given 300mg extract (5% hyperforin), mean plasma concentration (Cmax) of hyperforin was 150ng/mL, 3.5 hours after oral administration.13
  • The maximum plasma concentration was reached after 3.5 hours after oral administration of 300mg of Hypericum.13
  • After single dose intake of 612mg dry extract of St. John's wort, the maximum plasma concentration of hypericin was 3.14ng/mL, and time to maximum concentration was 8.1 hours; for pseudohypericin the maximum plasma concentration was 8.50ng/mL with a time to maximum concentration of 3.0 hours; and for hyperforin, the maximum plasma concentration was 83.5ng/mL with a time to maximum concentration of 4.4 hours.77 Quercetin and isorhamnetin showed two peaks of maximum plasma concentration separated by about 4 hours. The maximum plasma concentration of quercetin was 47.7ng/mL and the time to maximum concentration was 1.17 hours; for isorhamnetin, the maximum plasma concentration was 7.6ng/mL, and the time to maximum concentration was 1.53 hours.
  • Area under the curve (AUC): After a single dose intake of 612mg dry extract of St. John's wort, the AUC(0-infinity) for Hypericum was 75.96 hours x ng/mL; for pseudohypericin, the AUC(0-infinity) was 93.03 hours x ng/mL; for hyperforin, AUC(0-max) was 1009.0 hours x ng/mL; for quercetin, the AUC(0-infinity) was 318.7 hours x ng/mL; and for isorhamnetin, the AUC(0-infinity) was 98.0 hours x ng/mL.
  • Administering 900 or 1,200mg of Hypericum extract resulted in lower maximum concentration and AUC values than those expected.13
  • Bioavailability: Systemic availability of hypericin and pseudohypericin after oral administration has been estimated at 14% and 21%, respectively.78
  • Distribution: A distribution half-life of three hours was noted for hyperforin.13
  • Metabolism: Based on animal study, hyperforin is metabolized in the liver by the hydroxylation pathway.79 Four major Phase I metabolites, including 19-hydroxyhyperforin, 24-hydroxyhyperforin, 29-hydroxyhyperforin, and 34-hydroxyhyperforin, were isolated.79
  • Cytochrome P450: St. John's wort is an inducer of cytochrome P450 enzymes, particularly the CYP3A4 family.9 Moore et al. demonstrated that St. John's wort (hyperforin) activates a regulator (pregnane X receptor) of CYP3A4 transcription and thereby induces expression of 3A4 in human liver cells.10 Bray et al. demonstrated induction of 3A and 2E1 in mice.7 Upon review, it was noted that St. John's wort may inhibit CYP3A4 acutely and then induce this enzyme with repeated administration.80,8 CYP2C9, 2D6, and 3A4 inhibition has also been reported in in vivo and in vitro study.5,6 A brief, three-day trial did not demonstrate a similar significant effect on 2D6 or 3A4 enzymes.33
  • P-glycoprotein: St. John's wort is known to induce cytochrome P450 (CYP) 3A4 and P-glycoprotein through pregnane X-receptor activation. It has been suggested that this may be accomplished through increased multidrug resistant receptor 1 (MDR1) messenger ribonucleic acid (mRNA) and increased P-glycoprotein levels in the duodenal mucosa, possibly influenced by the MDR1 genotype.81,32,8,12
  • Elimination half-life: The elimination half-life ranges from 24-36 hours for hypericin40; between 9-19.64 hours for hyperforin77; between 9 and 23.76 hours for hypericin77,13; 25.39 hours for pseudohypericin77; 4.16 hours for quercetin77; and 4.45 hours for isorhamnetin77.
References
  1. Avato, P., Raffo, F., Guglielmi, G., Vitali, C., and Rosato, A. Extracts from St John's Wort and their antimicrobial activity. Phytother Res 2004;18(3):230-232. 15103670
  2. El Sherbiny, D. A., Khalifa, A. E., Attia, A. S., and Eldenshary, Eel. Hypericum perforatum extract demonstrates antioxidant properties against elevated rat brain oxidative status induced by amnestic dose of scopolamine. Pharmacol Biochem Behav 2003;76(3-4):525-533. 14643852
  3. Hunt, E. J., Lester, C. E., Lester, E. A., and Tackett, R. L. Effect of St. John's wort on free radical production. Life Sci 6-1-2001;69(2):181-190. 11441908
  4. Couldwell, W. T., Gopalakrishna, R., Hinton, D. R., He, S., Weiss, M. H., Law, R. E., Apuzzo, M. L., and Law, R. E. Hypericin: a potential antiglioma therapy. Neurosurgery 1994;35(4):705-710. 7808614
  5. Budzinski, J. W., Foster, B. C., Vandenhoek, S., and Arnason, J. T. An in vitro evaluation of human cytochrome P450 3A4 inhibition by selected commercial herbal extracts and tinctures. Phytomedicine 2000;7(4):273-282. 10969720
  6. Obach, R. S. Inhibition of human cytochrome P450 enzymes by constituents of St. John's Wort, an herbal preparation used in the treatment of depression. J Pharmacol Exp Ther 2000;294(1):88-95. 10871299
  7. Bray BJ, Perry NB, Menkes DB, and et al. St. John's wort extract induces CYP3A and CYP2E1 in the Swiss webster mouse. Toxicol Sci 2002;66(1):27-33.
  8. Cott JM. Herb-drug interactions. CNS Spectrums: The International Journal of Neuropsychiatric Medicine 2001;6:827-832.
  9. Roby, C. A., Anderson, G. D., Kantor, E., Dryer, D. A., and Burstein, A. H. St John's Wort: effect on CYP3A4 activity. Clin Pharmacol Ther 2000;67(5):451-457. 10824623
  10. Moore, L. B., Goodwin, B., Jones, S. A., Wisely, G. B., Serabjit-Singh, C. J., Willson, T. M., Collins, J. L., and Kliewer, S. A. St. John's wort induces hepatic drug metabolism through activation of the pregnane X receptor. Proc Natl Acad Sci USA 6-20-2000;97(13):7500-7502. 10852961
  11. Krusekopf, S. and Roots, I. St. John's wort and its constituent hyperforin concordantly regulate expression of genes encoding enzymes involved in basic cellular pathways. Pharmacogenet Genomics 2005;15(11):817-829. 16220113
  12. Turton-Weeks, SM, Barone GW, Gurley BJ, and et al. St. John's wort: a hidden risk for transplant patients. Prog Transplant 2001;11(2):116-120.
  13. Biber, A., Fischer, H., Romer, A., and Chatterjee, S. S. Oral bioavailability of hyperforin from hypericum extracts in rats and human volunteers. Pharmacopsychiatry 1998;31 Suppl 1:36-43. 9684946
  14. Cott, J. M. In vitro receptor binding and enzyme inhibition by Hypericum perforatum extract. Pharmacopsychiatry 1997;30 Suppl 2:108-112. 9342770
  15. Hansen, R. S., Paulsen, I., and Davies, M. Determinants of amentoflavone interaction at the GABA(A) receptor. Eur J Pharmacol 9-20-2005;519(3):199-207. 16129428
  16. Suzuki, O., Katsumata, Y., Oya, M., Bladt, S., and Wagner, H. Inhibition of monoamine oxidase by hypericin. Planta Med 1984;50(3):272-274. 6484033
  17. Bladt, S. and Wagner, H. Inhibition of MAO by fractions and constituents of hypericum extract. J Geriatr Psychiatry Neurol 1994;7 Suppl 1:S57-S59. 7857511
  18. Singer, A., Wonnemann, M., and Muller, W. E. Hyperforin, a major antidepressant constituent of St. John's Wort, inhibits serotonin uptake by elevating free intracellular Na+1. J Pharmacol Exp Ther 1999;290(3):1363-1368. 10454515
  19. Demisch L, Holzl J, Gollnik B, and et al. Identification of selective MAO-type-A inhibitors in Hypericum perforatum L. (Hyperforat). Pharmacopsychiat 1989;22:194.
  20. Thiede, H. M. and Walper, A. Inhibition of MAO and COMT by hypericum extracts and hypericin. J Geriatr Psychiatry Neurol 1994;7 Suppl 1:S54-S56. 7857510
  21. Schempp, C. M., Winghofer, B., Ludtke, R., Simon-Haarhaus, B., Schopf, E., and Simon, J. C. Topical application of St John's wort (Hypericum perforatum L.) and of its metabolite hyperforin inhibits the allostimulatory capacity of epidermal cells. Br J Dermatol 2000;142(5):979-984. 10809859
  22. Meruelo, D., Lavie, G., and Lavie, D. Therapeutic agents with dramatic antiretroviral activity and little toxicity at effective doses: aromatic polycyclic diones hypericin and pseudohypericin. Proc Natl Acad Sci USA 1988;85(14):5230-5234. 2839837
  23. Lavie, G., Valentine, F., Levin, B., Mazur, Y., Gallo, G., Lavie, D., Weiner, D., and Meruelo, D. Studies of the mechanisms of action of the antiretroviral agents hypericin and pseudohypericin. Proc Natl Acad Sci USA 1989;86(15):5963-5967. 2548193
  24. Schinazi, R. F., Chu, C. K., Babu, J. R., Oswald, B. J., Saalmann, V., Cannon, D. L., Eriksson, B. F., and Nasr, M. Anthraquinones as a new class of antiviral agents against human immunodeficiency virus. Antiviral Res 1990;13(5):265-272. 1697740
  25. Tang, J., Colacino, J. M., Larsen, S. H., and Spitzer, W. Virucidal activity of hypericin against enveloped and non-enveloped DNA and RNA viruses. Antiviral Res 1990;13(6):313-325. 1699494
  26. Wood S, Huffman J, Weber N, and et al. Antiviral activity of naturally occurring anthraquinones and anthraquinone derivatives. Planta Med 1990;56:651-652.
  27. Carpenter, S. and Kraus, G. A. Photosensitization is required for inactivation of equine infectious anemia virus by hypericin. Photochem Photobiol 1991;53(2):169-174. 1707176
  28. Hudson, J. B., Lopez-Bazzocchi, I., and Towers, G. H. Antiviral activities of hypericin. Antiviral Res 1991;15(2):101-112. 1650164
  29. Lopez-Bazzocchi, I., Hudson, J. B., and Towers, G. H. Antiviral activity of the photoactive plant pigment hypericin. Photochem Photobiol 1991;54(1):95-98. 1658826
  30. Degar, S., Prince, A. M., Pascual, D., Lavie, G., Levin, B., Mazur, Y., Lavie, D., Ehrlich, L. S., Carter, C., and Meruelo, D. Inactivation of the human immunodeficiency virus by hypericin: evidence for photochemical alterations of p24 and a block in uncoating. AIDS Research and Human Retroviruses 1992;8(11):1929-1936. 1283309
  31. Cohen, P. A., Hudson, J. B., and Towers, G. H. Antiviral activities of anthraquinones, bianthrones and hypericin derivatives from lichens. Experientia 2-15-1996;52(3):180-183. 8608821
  32. Durr, D., Stieger, B., Kullak-Ublick, G. A., Rentsch, K. M., Steinert, H. C., Meier, P. J., and Fattinger, K. St John's Wort induces intestinal P-glycoprotein/MDR1 and intestinal and hepatic CYP3A4. Clin Pharmacol Ther 2000;68(6):598-604. 11180019
  33. Markowitz, J. S., DeVane, C. L., Boulton, D. W., Carson, S. W., Nahas, Z., and Risch, S. C. Effect of St. John's wort (Hypericum perforatum) on cytochrome P-450 2D6 and 3A4 activity in healthy volunteers. Life Sci 1-21-2000;66(9):L133-L139. 10698361
  34. Müller WE, Rolli, M., Schafer, C., and Hafner, U. Effects of hypericum extract (LI 160) in biochemical models of antidepressant activity. Pharmacopsychiatry 1997;30 Suppl 2:102-107. 9342769
  35. Chatterjee SS, Bhattacharya SK, Singer A, and et al. Hyperforin inhibits synaptosomal uptake of neurotransmitters in vitro and shows antidepressant activity in vivo. Pharmazie 1998;53(3):9.
  36. Neary, J. T. and Bu, Y. Hypericum LI 160 inhibits uptake of serotonin and norepinephrine in astrocytes. Brain Res 1-23-1999;816(2):358-363. 9878826
  37. Franklin, M. Sub-chronic treatment effects of an extract of Hypericum perforatum (St. John's Wort, Li 160) on neuroendocrine responses to the 5-T2A agonist, DOI in the rat. Pharmacopsychiatry 2003;36(4):161-164. 12905103
  38. Staffeldt, B., Kerb, R., Brockmoller, J., Ploch, M., and Roots, I. Pharmacokinetics of hypericin and pseudohypericin after oral intake of the hypericum perforatum extract LI 160 in healthy volunteers. J Geriatr Psychiatry Neurol 1994;7 Suppl 1:S47-S53. 7857509
  39. Pietta, P., Gardana, C., and Pietta, A. Comparative evaluation of St. John's wort from different Italian regions. Farmaco 2001;56(5-7):491-496. 11482784
  40. Bombardelli E and Morazzoni P. Hypericum perforatum. Fitoterapia 1995;66(1):43-68.
  41. Nahrstedt, A. and Butterweck, V. Biologically active and other chemical constituents of the herb of Hypericum perforatum L. Pharmacopsychiatry 1997;30 Suppl 2:129-134. 9342774
  42. los Reyes, G. C. and Koda, R. T. Development of a simple, rapid and reproducible HPLC assay for the simultaneous determination of hypericins and stabilized hyperforin in commercial St. John's wort preparations. J Pharm Biomed Anal 2001;26(5-6):959-965. 11600308
  43. Jensen, A. G. and Hansen, S. H. Separation of hypericins and hyperforins in extracts of Hypericum perforatum L. using non-aqueous capillary electrophoresis with reversed electro-osmotic flow. J Pharm Biomed Anal 1-1-2002;27(1-2):167-176. 11682223
  44. Butterweck, V., Wall, A., Lieflander-Wulf, U., Winterhoff, H., and Nahrstedt, A. Effects of the total extract and fractions of Hypericum perforatum in animal assays for antidepressant activity. Pharmacopsychiatry 1997;30 Suppl 2:117-124. 9342772
  45. Bhattacharya, S. K., Chakrabarti, A., and Chatterjee, S. S. Activity profiles of two hyperforin-containing hypericum extracts in behavioral models. Pharmacopsychiatry 1998;31 Suppl 1:22-29. 9684944
  46. Calapai, G., Crupi, A., Firenzuoli, F., Inferrera, G., Squadrito, F., Parisi, A., De Sarro, G., and Caputi, A. Serotonin, norepinephrine and dopamine involvement in the antidepressant action of hypericum perforatum. Pharmacopsychiatry 2001;34(2):45-49. 11302563
  47. Di, Matteo, V, Di Giovanni, G., Di Mascio, M., and Esposito, E. Effect of acute administration of hypericum perforatum-CO2 extract on dopamine and serotonin release in the rat central nervous system. Pharmacopsychiatry 2000;33(1):14-18. 10721879
  48. Simbrey, K., Winterhoff, H., and Butterweck, V. Extracts of St. John's wort and various constituents affect beta-adrenergic binding in rat frontal cortex. Life Sci 1-9-2004;74(8):1027-1038. 14672758
  49. Wonnemann, M., Singer, A., and Muller, W. E. Inhibition of synaptosomal uptake of 3H-L-glutamate and 3H-GABA by hyperforin, a major constituent of St. John's Wort: the role of amiloride sensitive sodium conductive pathways. Neuropsychopharmacology 2000;23(2):188-197. 10882845
  50. Nathan, P. J. Hypericum perforatum (St John's Wort): a non-selective reuptake inhibitor? A reof the recent advances in its pharmacology. J Psychopharmacol. 2001;15(1):47-54. 11277608
  51. Bennett, D. A., Jr., Phun, L., Polk, J. F., Voglino, S. A., Zlotnik, V., and Raffa, R. B. Neuropharmacology of St. John's Wort (Hypericum). The Annals of Pharmacotherapy 1998;32(11):1201-1208. 9825087
  52. Hostanska, K., Reichling, J., Bommer, S., Weber, M., and Saller, R. Hyperforin a constituent of St John's wort (Hypericum perforatum L.) extract induces apoptosis by triggering activation of caspases and with hypericin synergistically exerts cytotoxicity towards human malignant cell lines. Eur J Pharm Biopharm 2003;56(1):121-132. 12837490
  53. Di Carlo, G., Nuzzo, I., Capasso, R., Sanges, M. R., Galdiero, E., Capasso, F., and Carratelli, C. R. Modulation of apoptosis in mice treated with Echinacea and St. John's wort. Pharmacol Res 2003;48(3):273-277. 12860446
  54. Panossian AG, Gabrielian E, Manvelian V, and et al. Immunosuppressive effects of hypericin on stimulated human leukocytes: inhibition of the arachidonic acid release, leukotriene B4and Interleukin-1alpha production, and activation of nitric oxide formation. Phytomedicine 1996;3(1):19-28.
  55. Thiele, B., Brink, I., and Ploch, M. Modulation of cytokine expression by hypericum extract. J Geriatr Psychiatry Neurol 1994;7 Suppl 1:S60-S62. 7857513
  56. Zhou, C., Tabb, M. M., Sadatrafiei, A., Grun, F., Sun, A., and Blumberg, B. Hyperforin, the active component of St. John's wort, induces IL-8 expression in human intestinal epithelial cells via a MAPK-dependent, NF-kappaB-independent pathway. J Clin Immunol 2004;24(6):623-636. 15622447
  57. Winkler, C., Wirleitner, B., Schroecksnadel, K., Schennach, H., and Fuchs, D. St. John's wort (Hypericum perforatum) counteracts cytokine-induced tryptophan catabolism in vitro. Biol Chem 2004;385(12):1197-1202. 15653434
  58. Dona, M., Dell'Aica, I., Pezzato, E., Sartor, L., Calabrese, F., Della, Barbera M., Donella-Deana, A., Appendino, G., Borsarini, A., Caniato, R., and Garbisa, S. Hyperforin inhibits cancer invasion and metastasis. Cancer Res 9-1-2004;64(17):6225-6232. 15342408
  59. Feisst, C. and Werz, O. Suppression of receptor-mediated Ca2+ mobilization and functional leukocyte responses by hyperforin. Biochem Pharmacol 4-15-2004;67(8):1531-1539. 15041470
  60. Gobbi, M., Moia, M., Funicello, M., Riva, A., Morazzoni, P., and Mennini, T. In vitro effects of the dicyclohexylammonium salt of hyperforin on interleukin-6 release in different experimental models. Planta Med 2004;70(7):680-682. 15303261
  61. Pajonk, F., Scholber, J., and Fiebich, B. Hypericin-an inhibitor of proteasome function. Cancer Chemother Pharmacol 2005;55(5):439-446. 15672261
  62. Hudson, J. B., Graham, E. A., and Towers, G. H. Antiviral assays on phytochemicals: the influence of reaction parameters. Planta Med 1994;60(4):329-332. 7938267
  63. Dimpfel, W., Todorova, A., and Vonderheid-Guth, B. Pharmacodynamic properties of St. John's wort-A single blind neurophysiological study in healthy subjects comparing two commercial preparations. Eur J Med Res 8-25-1999;4(8):303-312. 10471542
  64. Johnson, D., Ksciuk, H., Woelk, H., Sauerwein-Giese, E., and Frauendorf, A. Effects of hypericum extract LI 160 compared with maprotiline on resting EEG and evoked potentials in 24 volunteers. J Geriatr Psychiatry Neurol 1994;7 Suppl 1:S44-S46. 7857508
  65. Schellenberg, R., Sauer, S., and Dimpfel, W. Pharmacodynamic effects of two different hypericum extracts in healthy volunteers measured by quantitative EEG. Pharmacopsychiatry 1998;31 Suppl 1:44-53. 9684947
  66. Schulz, H. and Jobert, M. Effects of hypericum extract on the sleep EEG in older volunteers. J Geriatr Psychiatry Neurol 1994;7 Suppl 1:S39-S43. 7857507
  67. Sharpley, A. L., McGavin, C. L., Whale, R., and Cowen, P. J. Antidepressant-like effect of Hypericum perforatum (St John's wort) on the sleep polysomnogram. Psychopharmacology (Berl) 1998;139(3):286-287. 9784087
  68. Sharpley AL, McGavin Cl, Whale R, and Cowen PJ. Hypericum perforatum (St. John's Wort) and depression?: Changes in sleep architecture in normal volunteers. XXIst Collegium Internationale Neuro-psychopharmacologicum, Glasgow, Scotland 12-16th July, 1998.
  69. Chatterjee, S. S., Bhattacharya, S. K., Wonnemann, M., Singer, A., and Muller, W. E. Hyperforin as a possible antidepressant component of hypericum extracts. Life Sci 1998;63(6):499-510. 9718074
  70. Teufel-Mayer, R. and Gleitz, J. Effects of long-term administration of hypericum extracts on the affinity and density of the central serotonergic 5-HT1 A and 5-HT2 A receptors. Pharmacopsychiatry 1997;30 Suppl 2:113-116. 9342771
  71. Schule, C., Baghai, T., Ferrera, A., and Laakmann, G. Neuroendocrine effects of Hypericum extract WS 5570 in 12 healthy male volunteers. Pharmacopsychiatry 2001;34 Suppl 1:S127-S133. 11518061
  72. Yu, P. H. Effect of the Hypericum perforatum extract on serotonin turnover in the mouse brain. Pharmacopsychiatry 2000;33(2):60-65. 10761821
  73. Jensen, A. G., Hansen, S. H., and Nielsen, E. O. Adhyperforin as a contributor to the effect of Hypericum perforatum L. in biochemical models of antidepressant activity. Life Sci 2-23-2001;68(14):1593-1605. 11263672
  74. Raffa, R. B. Screen of receptor and uptake-site activity of hypericin component of St. John's wort reveals sigma receptor binding. Life Sci 1998;62(16):L265-L270. 9585174
  75. Schroeder, C., Tank, J., Goldstein, D. S., Stoeter, M., Haertter, S., Luft, F. C., and Jordan, J. Influence of St John's wort on catecholamine turnover and cardiovascular regulation in humans. Clin Pharmacol Ther 2004;76(5):480-489. 15536462
  76. Wong, M. L., O'Kirwan, F., Hannestad, J. P., Irizarry, K. J., Elashoff, D., and Licinio, J. St John's wort and imipramine-induced gene expression profiles identify cellular functions relevant to antidepressant action and novel pharmacogenetic candidates for the phenotype of antidepressant treatment response. Mol Psychiatry 1-13-2004;14743185
  77. Schulz, H. U., Schurer, M., Bassler, D., and Weiser, D. Investigation of the bioavailability of hypericin, pseudohypericin, hyperforin and the flavonoids quercetin and isorhamnetin following single and multiple oral dosing of a hypericum extract containing tablet. Arzneimittelforschung 2005;55(1):15-22. 15727160
  78. Kerb, R., Brockmoller, J., Staffeldt, B., Ploch, M., and Roots, I. Single-dose and steady-state pharmacokinetics of hypericin and pseudohypericin. Antimicrob Agents Chemother 1996;40(9):2087-2093. 8878586
  79. Cui, Y., Ang, C. Y., Beger, R. D., Heinze, T. M., Hu, L., and Leakey, J. In vitro metabolism of hyperforin in rat liver microsomal systems. Drug Metab Dispos 2004;32(1):28-34. 14709617
  80. Cott, J. M., Rosenthal, N., and Blumenthal, M. St John's wort and major depression. JAMA 7-4-2001;286(1):42-45. 11434815
  81. Schwarz, U. I., Hanso, H., Oertel, R., Miehlke, S., Kuhlisch, E., Glaeser, H., Hitzl, M., Dresser, G. K., Kim, R. B., and Kirch, W. Induction of intestinal P-glycoprotein by St John's wort reduces the oral bioavailability of talinolol. Clin Pharmacol Ther 2007;81(5):669-678. 17392718
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler
back to top