Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Key Publications and Guidance on Extracellular Vesicle Preparation
Read This Before You Start Your Extracellular Vesicle Research!
EV preparations can vary in molecular composition and function, depending on:
  • Cell culture viability
  • Apoptotic index
  • Cell proliferation rate
  • Status of the cell culture medium
  • Handling conditions
  • Other experimental parameters
To support EV researchers trying to navigate this rapidly changing field, we've made available to you a suite of resources for your easy reference.
  1. Wall Poster: Preparing functional extracellular vesicles (EVs): Join the rEVolution.
    Hang this unique poster in your lab to be reminded of EV biogenesis pathways, cell type-specific EV processes, and characteristic protein and miRNA markers.
    Download Poster
  2. Review article in BioProcessing Journal (in press): Preparing biotherapeutic extracellular vesicles: ultrafilters, MSCs and the regulatory horizon.
  3. Detection of internalized exosomes in single cancer cells using Amnis® imaging flow cytometry: View Poster Presented at CYTO 2013
    Exosomes were co-cultured with tumor cells and analyzed for internalization of PKH26+ particles in the tumor cell lines using the Amnis ImageStream®X imaging flow cytometer.
ISEV Guidances: Quick Links
The International Society for Extracellular Vesicles (ISEV) has published guidances for preparation and analysis, recognizing that the exact methods could depend on the scientific question being asked.
Read the most recent guidances:
Witwer KW et al. Standardization of sample collection, isolation and analysis methods in extracellular vesicle research. 2013 May 27;2.
Lötvall J et al. Minimal experimental requirements for definition of extracellular vesicles and their functions: a position statement from the International Society for Extracellular Vesicles. 2014 Dec 22;3:26913.