Centrifugal Filters for Non-Clinical Samples
Whether you need to analyze DNA from forensic samples or separate low-molecular-weight solutes from fermentation broths and cell lysates, we provide specialized centrifugal filters that match your application. Centrifugal filters optimize recovery of DNA and protein from biological samples
  Centrifugal filters optimize recovery
of DNA and protein from biological
samples
Application Sample Volume Range Device
Concentration of genomic DNA and protein ≤0.5 mL Microcon® DNA Fast Flow Centrifugal Filters
Microcon®10K and 30K MWCO Filters
Clarification and filtration of aqueous and some solvent-based samples ≤0.5 mL Ultrafree®-MC Centrifugal Filters
Ultrafree®-CL Centrifugal Filters
High-solute sample concentration and/or desalting ≤15 mL Centriprep® Centrifugal Filters