Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Ultrafree®-MC and Ultrafree-CL Centrifugal Filters with Microporous Membrane
Ultrafree®-MC and -CL centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions. These devices are available in two processing volumes with a range of microporous membranes.
 Advantages of Ultrafree®-MC Filters with Microporous Membrane
  • Five different pore sizes from 0.1 to 5.0 µm
  • Pre-sterilized units also available
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 1.5 mL tubes
Order Ultrafree®-MC Centrifugal filters
Centrifuge and Rotor Recommendations
 Advantages of Ultrafree®-CL Filters with Microporous Membrane
  • High recovery Durapore® (PVDF) and hydrophilic PTFE membranes
  • Five different pore sizes from 0.1 to 5.0 µm
  • Pre-sterilized units also available
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 15 mL tubes
Order Ultrafree®-CL Centrifugal filters
Sterile Ultrafree®-MC and CL Centrifugal Filter Units with Microporous Membrane
  • Easy, pre-sterilized, centrifugal sample clarification units for either 0.5 mL (MC) or 2 mL (CL) maximum volumes.
  • High recovery Durapore® (PVDF) membrane
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 1.5 mL tubes (MC) or 15 mL tubes (CL)
Order Ultrafree®-MC and CL Sterile Centrifugal Filters
 Applications
  • Removal of particles and precipitates from aqueous and some solvent-based samples
  • Ideal for use with protein and nucleic acid solutions