Insect Cell Lysis
Insect PopCulture® Reagent is a detergent-based lysis reagent specifically formulated for total insect cell culture (in suspension or adherent) extraction without the need for centrifugation.
CytoBuster™ Protein Extraction Reagent is a proprietary formulation of detergents optimized for fast, efficient extraction of soluble proteins from mammalian and insect cell to be used in activity assays.
PhosphoSafe™ Extraction Buffer is a detergent and phosphatase inhibitor mixture optimized for fast, efficient extraction of soluble proteins from mammalian and insect cell preserving the phosphorylation state of your sample.
Purification of His•Tag® β-galactosidase from baculovirus-infected insect cell cultures.