Mammalian Cell Lysis
Choose a mammalian cell lysis reagent based on your sample and downstream detection methods. MilliporeSigma’s lysis reagents support preparation of mammalian and insect proteins, including cytosolic, nuclear and phosphorylated proteins.
CytoBuster™ Protein Extraction Reagent is a proprietary formulation of detergents optimized for fast, efficient extraction of soluble proteins from mammalian and insect cells to be used in activity assays.
The PhosphoSafe™ Extraction Reagent is a detergent and phosphatase inhibitor mixture optimized for fast, efficient extraction of soluble proteins from mammalian and insect cells and preserves the phosphorylation state of extracted proteins.
ProteoExtract Kits – dedicated extraction kits from mammalian cells and tissues.
Research Poster:
Optimizing Extraction of β-amyloid from Brain Tissue
Click image to enlarge.
In Direct Extraction, CytoBuster™ reagent (“Cyto”), TBS and Triton® X-114 appeared to be more efficient in solubilizing Aβ 1-40 fragment than Formic Acid (FA).
Presented at Neuroscience 2015 Conference
Download Now