Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Rotor Guide - Centricon® Plus-70
  • 62x123 mm
  • Maximum g-force 3500 x g
  • Swinging bucket rotor
Learn more about Centricon® Plus-70
 Select Manufacturer to See Recommendations
Note: Centrifuges and rotors listed are not available from us. Please confirm product availabilty with the manufacturer.
Beckman
Centrifuge Model Rotor Adapter Required
Allegra X12 SX4750 349946
Allegra X12 SX4750A 349946
5810 / 5810R A-4-62 22638441
back to top
Hettich
Centrifuge Model Rotor Adapter Required
ROTANTA460/460R 5624 5620/4841
ROTANTA460/460R 4446 5092/6319
ROTINA35/35R 1724 1790/6319
back to top
Kendro
Centrifuge Model Rotor Adapter Required
Legend Mach 1.6, Multifuge 1/1R 75002002 75007622 One per adapter
Legend T/RT, Multifuge 3/3R 75006441 75008144 One per adapter
Legend T/RT, Multifuge 3/3R 75006446 75008144 One per adapter
back to top
Thermo/Forma
Centrifuge Model Rotor Adapter Required
5530/5532 1L GP/GPR 5800245, 5800268 (without sealing cover), (5850968 - for use with rotors listed to left)
5530/5532 1L GP/GPR 5800224, 5852242 (without sealing cover), (5850968 - for use with rotors listed to left)
5677/5678 1L MP/MPR 5800224 (without sealing cover)
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800218 5805780
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800228 5805780
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800216 5805780
5694/5696 1L HP/HPR 5808947 5806558
back to top
Thermo/IEC
Centrifuge Model Rotor Adapter Required
HN-SII, Centra® HN 245, 268 (without sealing cover)
Centra® CL4 224, 52242 (without sealing cover)
Centra CL3/CL3R 243 6558E
Centra CL3/CL3R 245, 268 (without sealing cover) 50968 - for use with rotors listed to left)
Centra CL3/CL3R 224, 52242 (without sealing cover) (50968 - for use with rotors listed to left)
Centra MP4/MP4R 224 (without sealing cover)
Centra 7/7R 210, 216 5780
Centra 8/8R 210, 216 5780
Centra GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 218A 5780
Centra GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 228 5780
Centra GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 216 5780
Multi/ Multi RF 8947 6558E
PR 7000M 219A, 256, 949, 966, 981 5780
Model K, EXD 219A, 256 5780
Model K, EXD 949 N/N
Model K, EXD 52592 N/N
back to top