Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Rotor Guide - Centriprep®
  • 28.1x135 mm
  • Centriprep®-3 and –10, 3000 x g; Centriprep-30 and -50, 1500 x g
  • Use any variable speed centrifuge up to 3000 x g with swinging bucket rotor or fixed angle rotor that can accommodate 50 ml tubes.
Centriprep® centrifugal filter
 Select Manufacturer to See Recommendations
Note: Centrifuges and rotors listed are not available from . Please confirm product availabilty with the manufacturer.
Beckman
Centrifuge Model Rotor Adapter Required
Allegra X12 SX4750 359474
Allegra X12 SX4750A 359474
back to top
Eppendorf
Centrifuge Model Rotor Adapter Required
5810 / 5810R A-4-62 22638386
back to top
Hermle    
Centrifuge Model Rotor Adapter Required
C0200-A C0200-97  
back to top
Hettich
Centrifuge Model Rotor Adapter Required
ROTANTA460/460R 5624 5625/4740
ROTANTA460/460R 5624 5620/4840
ROTANTA460/460R 5615 1446
ROTINA35/35R 1792 1446
ROTINA35/35R 1720 1446
back to top
Kendro
Centrifuge Model Rotor Adapter Required
Biofuge Primo/Primo R 75007588 75003102 (Without rotor lid up to 3000 xg, 3 filters only)
Biofuge Stratos 75003057 75003102 (Without rotor lid up to 3000 xg, 3 filters only)
Biofuge Stratos 75008172 75008182
Legend Mach 1.6, Multifuge 1/1R 75002006 75003102 (Without rotor lid up to 3000 xg, 3 filters only)
Legend Mach 1.6, Multifuge 1/1R 75002002 75007574
Legend Mach 1.6, Multifuge 1/1R 75002001 75005324
Legend T/RT, Multifuge 3/3R 75003057 75003102 (Without rotor lid up to 3000 xg, 3 filters only)
Legend T/RT, Multifuge 3/3R 75006441 75006493
Legend T/RT, Multifuge 3/3R 75006446 75006491
back to top
Thermo/Forma
Centrifuge Model Rotor Adapter Required
5528 400ML GP 5858012  
5529 400ML GP 5800215* 5800325/5800320
5530 400ML GP 5800215* 5800326/5800320
5530/5532 1L GP/GPR 5800215* 5800325/5800320
5530/5532 1L GP/GPR 5800215* 5800326/5800320
5530/5532 1L GP/GPR 5859581, 5858012, 5858162 (5850968 - for use with rotors listed to left)
5530/5532 1L GP/GPR 5800243 5806556
5677/5678 1L MP/MPR 5800215* 5800325/5800320
5677/5678 1L MP/MPR 5800215* 5800326/5800320
5677/5678 1L MP/MPR 5858012, 5858162  
5540/5542 1L MP/MPR 5806555 5806556
5540/5542 1L MP/MPR 5807685 5806650
5694/5696 1L HP/HPR 5808685 5806650
5694/5696 1L HP/HPR 5808850  
5694/5696 1L HP/HPR 5808947 5806556
5660/5662 3L 2500 GP/GPR 5806554 5807007 (sealed), 5805707 (unsealed)
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800216, 5800228 5805707
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800218 5805707
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800825 (without sealing cover)
back to top
Thermo/IEC
Centrifuge Model Rotor Adapter Required
Clinical 58012  
Centra® CL2 58012  
Centra® CL2 215* 325/320
Centra® CL2 215* 326/320
HN-SII, Centra® HN 958; 59581; 325/320
HN-SII, Centra® HN 958; 59582 326/320
HN-SII, Centra® HN 245, 268 7231
HN-SII, Centra® HN 58012, 58162 N/N
Centra® CL4 215* 325/320
Centra® CL4 215* 326/320
Centra® CL4 224*, 52242* 7231
Centra® CL4 58012, 58162 N/N
Centra® CL3/CL3R 215* 325/320
Centra® CL3/CL3R 215* 326/320
Centra® CL3/CL3R 59581, 58012, 58162 (50968 - for use with rotors listed to left)
Centra® CL3/CL3R 243 6556E
Centra® MP4/MP4R 215* 325/320
Centra® MP4/MP4R 215* 326/320
Centra® MP4/MP4R 58012, 58162 N/N
Centra® CL5/CL5R 6555C 6556E
Centra® CL5/CL5R 7685C 6650E
Multi/ Multi RF 8685 6650E
Multi/ Multi RF 8850 N/N
Multi/ Multi RF 8947 6556E
Centra® 7/7R 210, 216 7007E (sealed), 5707 (unsealed)
Centra® 7/7R 825S (without sealing cover)
Centra® 8/8R 210, 216 7007E (sealed), 5707 (unsealed)
Centra® 8/8R 825S N/N (without sealing cover)
Centra® GP6/GP6R 6554C 7007E (sealed), 5707 (unsealed)
Centra® GP8/GP8R (Bench),GP8K/GP8RK (Kneewell),GP8F/GP8RF (Floor) 216, 228 5707
Centra® GP8/GP8R (Bench),GP8K/GP8RK (Kneewell),GP8F/GP8RF (Floor) 218A 5707
Centra® GP8/GP8R (Bench),GP8K/GP8RK (Kneewell),GP8F/GP8RF (Floor) 825S (without sealing cover)
C-6000, CR-6000 256, 219 5707
C-6000, CR-6000 284 384S/7231
C-6000, CR-6000 921, 269, 253 325/305
C-6000, CR-6000 921, 269, 253 325/320
C-6000, CR-6000 921, 269, 253 326/305
C-6000, CR-6000 921, 269, 253 326/320
Model K, EXD 219A, 256, 949 5707
Model K, EXD 284, 259 384S/7231
Model K, EXD 921, 269, 253 325/305
Model K, EXD 921, 269, 253 325/320
Model K, EXD 921, 269, 253 326/305
Model K, EXD 921, 269, 253 326/320
PR-7000M 219A, 256, 949, 966, 981 5707
PR-7000M 284, 259 384S/7231
PR-7000M 269, 253 325/305
PR-7000M 269, 253 325/320
PR-7000M 269, 253 326/305
PR-7000M 269, 253 326/320
B-22M 877, 963 7231
back to top