Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Rotor Guide - Ultrafree®-CL
  • 16.3x77 mm
  • Maximum g-force 5000 x g
  • Fixed-angle rotor
Centriprep® centrifugal filter
 Select Manufacturer to See Recommendations
Note: Centrifuges and rotors listed are not available from us. Please confirm product availabilty with the manufacturer.
Beckman    
Centrifuge Model Rotor Adapter Required
Allegra X12 FX6100 392824
back to top
Eppendorf    
Centrifuge Model Rotor Adapter Required
5810/5810R A-4-62 22638262
Max g's is 4,400
back to top
Hermle    
Centrifuge Model Rotor Adapter Required
C0323 C0252-85  
back to top
Hettich    
Centrifuge Model Rotor Adapter Required
EBA21 1116 1635 with 18-0716 black rubber inserts
MIKRO22/22R 1016 1635 with 18-0716 black rubber inserts
ROTANTA460/460R 5615 1451 with 18-0716 black rubber inserts
ROTINA35/35R 1792 1451 with 18 - 0716 black rubber inserts
ROTOFIX32 1620A 1451 with 18-0716 black rubber inserts
UNIVERSAL32/32R 1620A 1451 with 18-0716 black rubber inserts
back to top
Kendro    
Centrifuge Model Rotor Adapter Required
Biofuge Primo/Primo R 75007588 76002906
Biofuge Stratos 75003057 76002906
Biofuge Stratos 75003336  
Biofuge Stratos 75003335 76002904
Biofuge Stratos 750003334 76002906
Legend Mach 1.6, Multifuge 1/1R 75002006 76002906
Legend Mach 1.6, Multifuge 1/1R 75002001 75002021
Legend Mach 1.6, Multifuge 1/1R 75002002 75007572
Legend T/RT, Multifuge 3/3R 75003057 76002906
Legend T/RT, Multifuge 3/3R 75006441 75006494
Legend T/RT, Multifuge 3/3R 75006446 75006492
back to top
Thermo/IEC    
Centrifuge Model Rotor Adapter Required
Clinical / Centra® CL2 809, 58092 302
Clinical / Centra® CL2 801, 58012 305/1106
Clinical / Centra® CL2 803 302
Clinical / Centra® CL2 804S 323/1106/571
HN-SII, Centra® HN 809, 58092 302
HN-SII, Centra® HN 801, 58012 305/1106
HN-SII, Centra® HN 815, 58153 302
HN-SII, Centra® HN 816, 58162 305/1106
HN-SII, Centra® HN 803 302
HN-SII, Centra® HN 804S 323/1106/571
Centra CL4 809, 58092 302
Centra CL4 801, 58012 305/1106
Centra CL4 815, 58153 302
Centra CL4 816, 58162 305/1106
Centra CL4 803 302
Centra CL4 804S 323/1106/571
Centra® CL3/CL3R 809, 58092 302, (50968 - for use with rotors listed to left)
Centra® CL3/CL3R 801, 58012 305/ 1106,(50968 - for use with rotors listed to left)
Centra® CL3/CL3R 803 (50968 - for use with rotors listed to left)
Centra® CL3/CL3R 804S 1106, (50968 - for use with rotors listed to left)
Centra® CL3/CL3R 816, 58162 305/1106
Centra® CL5/CL5R 7685C 6650E/1106
Centra® MP4/MP4R 809, 58092 302
Centra® MP4/MP4R 801, 58012 305/1106
Centra® MP4/MP4R 815, 58153 302
Centra® MP4/MP4R 816, 58162 305/1106
Centra® MP4/MP4R 803 302
Centra® MP4/MP4R 804S 323/1106/571
Multi/ Multi RF 8685 6650E/1106
Multi/ Multi RF 8850 50546/1106
back to top