Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Rotor Guide - Ultrafree®-MC
  • 10.6x45 mm
  • Maximum g-force 12000 x g
  • Fixed-angle rotor
Ultrafree®-MC Centrifugal Filter Unit
 Select Manufacturer to See Recommendations
Note: Centrifuges and rotors listed are not available from us. Please confirm product availabilty with the manufacturer.
Beckman
Centrifuge Model Rotor Adapter Required
Allegra 64R / Avanti 30 / Allegra 21/21R / GS-15/15R F2402H  
Microfuge 22R F301.5  
Microfuge 22R F241.5  
Microfuge 22R / Microfuge 18 F241.5P  
TJ-25 / Allegra 25R TA-15-1.5  
back to top
Eppendorf    
Centrifuge Model Rotor Adapter Required
5415D & R 22636502  
5417C & R F-45-30-11  
5417C & R FA-45-30-11  
5417C & R FA-45-24-11  
5804 / 5804R F-34-6-38 22637215
5810 / 5810R F-34-6-38 22637215
Minispin Included  
Minispin Plus Included  
back to top
Hermle
Centrifuge Model Rotor Adapter Required
Spectrafuge 16M or 24D    
back to top
Hettich
Centrifuge Model Rotor Adapter Required
EBA21 1112  
EBA21 1095  
EBA21 1089  
MIKRO200/200R 2424  
MIKRO200/200R 2430  
ROTINA35/35R 1789  
ROTANTA460/460R 4489  
UNIVERSAL32/32R 1612  
UNIVERSAL32/32R 1689  
MIKRO22/22R 1157  
MIKRO22/22R 1195  
MIKRO22/22R 1189  
MIKRO22/22R 1158  
back to top
Kendro
Centrifuge Model Rotor Adapter Required
Biofuge Pico/Fresco 75003328  
Labofuge 400/400R 75003325  
Legend Mach 1.6/1.6R, Multifuge 1/1R 75002006 76002905
Legend Mach 1.6/1.6R, Multifuge 1/1R 75003448  
Legend Mach 1.6/1.6R, Multifuge 1/1R 75002001 75005335
Legend Mach 1.6/1.6R, Multifuge 1/1R 75002002 75007578
Legend T/RT, Multifuge 3/3R 75003332  
Legend T/RT, Multifuge 3/3R 75003057 76002905
Legend T/RT, Multifuge 3/3R 75003334 76002905
Legend T/RT, Multifuge 3/3R 75006441 75008132
Legend T/RT, Multifuge 3/3R 75006446 75006452
back to top
Thermo/Forma
Centrifuge Model Rotor Adapter Required
5677/5678 1L MP/MPR 5800891, 5800851 5850036 hub adapter for 5800891, 5800851
5540/5542 1L MP/MPR 5807851  
5694/5696 1L HP/HPR 5808848  
5677/5678 1L MP/MPR 5800891, 5800851 5850036 hub adapter for 5800891, 5800851
5540/5542 1L MP/MPR 5807851  
back to top
Thermo/IEC
Centrifuge Model Rotor Adapter Required
Centra® MP4/ MP4R 891, 851 50036 hub adapter for 891, 851
Micromax/ Micromax RF 891, 851  
MicroLite/MicroLite RF 891, 851  
Multi/ Multi RF 8848  
back to top