Mass Spectrometry
Mass Spectrometry Calibration Standards
 Standardize your proteomics research
We offer a variety of mass spectrometry calibration standards for quantitative proteomics. These high quality products help to standardize your proteomics research and provide the best quality control for the precision and accuracy of your MS data.
Universal Proteomics Standard
UPS1
• A well-defined, well-characterized
  standard made up of 48 proteins from
  6-83 KD (5pmoles of each)
UPS2
• A well-defined, well-characterized
  standard made up of 6 mixtures of 8
  proteins, presenting a dynamic range of
  50 pmoles-500amoles
MS Qual/Quant QC Mix
MSQC1
• Contains 6 tryptically digested human
  proteins and 14 SIL peptides
• Assesses repeatability between runs,
  system stability, and inter- and intra-
  platform and lab comparisons
• Predigested and injection-ready
MS Retention Time Calibration Mix
MSRT1
• An injection-ready mixture containing
  14 stable isotope-labeled peptides
MSRT2
• An injection-ready mixture containing
  9 native proteins
MS Phosphomix  
Light Products
MSP1L
MSP2L
MSP3L
Heavy (SIL) Products
MSP1H
MSP2H
MSP3H
Related Products
SILu™MAb & SigmaMAb
SILu™Prot
SILu™PrEST
AQUA™
SOLu-Trypsin