Immobilon®-E Sample Request
We’ve launched our newest addition to the Immobilon® family: Immobilon®-E, the first PVDF blotting membrane that wets out in aqueous transfer buffer. Skip the alcohol pre-wet step! Rethink Western blotting and request your sample today.
Try our new Immobilon®-E transfer membrane for your next Western blotting experiment!
Fields indicated by an * are required.
You can unsubscribe at any time