Mesenchymal Stem Cell Culture
Human mesenchymal stem cells (MSCs) are multipotent adult stem cells, which have the capacity for multi-lineage differentiation, giving rise to a variety of mesenchymal phenotypes such as osteoblasts (bone), adipocytes (fat), and chondrocytes (cartilage). Mesenchymal stem cells usually reside in the bone marrow stroma, but have also been found in adipose, liver, spleen, peripheral blood, umbilical cord blood, and other mesenchymal tissues. Due to their capacity for self-renewal over long periods of time and the ability to differentiate into specialized cells, interest in understanding the biology of MSC cultures has increased, especially for their therapeutic potential for a variety of diseases. We offer a broad range of tools and technologies to culture mesenchymal stem cells including low passage MSC lines from various tissues, optimized expansion and differentiation media and a broad range or MSC related antibodies and cellular dyes.
 Mesenchymal stem cell overview. Trilineage differentiation of multipotent human mesenchymal stem cells (MSCs) includes differentiation into osteoblasts, adipocytes and chondrocytes.
Figure 1. Mesenchymal stem cell overview. Trilineage differentiation of multipotent human mesenchymal stem cells (MSCs) includes differentiation into osteoblasts, adipocytes and chondrocytes.
 Mesenchymal Stem Cell Lines
Ordering Info
Cell Type Brand Format Part Number
Human Bone-Marrow MSCs PromoCell* 5X105 Cryopreserved Cells C-12974
  Sigma 1X106 Cryopreserved Cells SCC034
  PromoCell* 5X105 Live Proliferating Cells C-12975
  PromoCell* 1X106 Cell Pellet C-14090
Human Adipose MSCs PromoCell* 5X105 Cryopreserved Cells C-12977
  Sigma 1X106 Cryopreserved Cells SCC038
  PromoCell* 5X105 Live Proliferating Cells C-12978
  PromoCell* 1X106 Cell Pellet C-14092
Human Umbilical Cord MSCs PromoCell* 5X105 Cryopreserved Cells C-12971
  PromoCell* 5X105 Live Proliferating Cells C-12972
  PromoCell* 1X106 Cell Pellet C-14091
Human ES Cell Derived MSCs Sigma 1X106 Cryopreserved Cells SCC036
Rat Bone-Marrow MSCs Sigma 1X106 Cryopreserved Cells SCR027
*Application Note: International Society for Cellular Therapy (ISCT) Guidelines on Multipotent Human Mesenchymal Stem Cell (MSC) Characterization.
Human mesenchymal stem cell markers.
Figure 2. Human mesenchymal stem cell markers. Human MSCs express H-CAM (CD44) (A), THY-1 (CD90) (B), and STRO-1 (C). Nuclei were visualized with DAPI (blue). Expression of hematopoietic stem cell markers CD19 and CD14 and endothelial marker CD146 were not observed (data not shown).
 Mesenchymal Stem Cell Media
MSC Expansion Media
We offer a variety of optimized and high performing complete mesenchymal stem cell expansion media. From low-serum to GMP compliant media options, we offer a solution to every type of MSC researcher.
Ordering Info
Serum Containing Defined/Xeno-Free GMP Grade
Mesenchymal Stem Cell Growth Medium 2 (PromoCell)
Mesenchymal Stem Cell Expansion Medium (Sigma)
Mesenchymal Stem Cell Growth Medium DXF (PromoCell)
Human Mesenchymal-XF Expansion Medium (Sigma)
Stemline® Mesenchymal Stem Cell Expansion Medium (Sigma)
Low FBS (2%) cell culture media for human mesenchymal stem cells. Complete xeno-free and serum-free media that is free of animal-derived components. Produced in a GMP state-of-the-art facility with an available DeviceMaster File (DMF).
Product Focus: PLTMax® Human Platelet Lysate
PLTMax Human Platelet LysateA Superior Alternative to FBS for Human MSC Cultures
With the increasing interest and utilization of human cellular therapy, PLTMax Human Platelet Lysate meets the need for a research grade cell culture supplement that is human derived, serum-free, and cost effective for human mesenchymal stem cell cultures.
Application Note: PLTMax® Human Platelet Lysate (hPL): A Superior Serum-Free Media Supplement for Robust Mesenchymal Stem Cell Expansion
Features and Benefits:
  • Increased cell growth kinetics vs. FBS supplemented media
  • Cost effective alternative to serum free media
  • Manufactured to reduce lot-to-lot variation
  • Stable supply and price
  • Human derived material applicable for translational research applications
PLTMax supports better growth of hMSCs than DMEM + FBS
Figure 3. PLTMax supports better growth of hMSCs than DMEM + FBS. A: Growth of hMSCs cultured in αMEM supplemented with 5 % HPL or 10 % FBS as compared to DMEM + 10 % FBS over 4 passages in T150 flasks, or B: in duplicate 125 mL spinner flasks f or 7 days, demonstrating enhanced growth in αMEM + HPL
Product No. Description
SCM141 PLTMax Human Platelet Lysate, 100ml
SCM142 PLTMax Human Platelet Lysate, 500ml
SCM151 PLTGold Human Platelet Lysate (Heparin-Free), 100ml
SCM152 PLTGold Human Platelet Lysate (Heparin-Free), 500ml
MSC Qualified Fetal Bovine Serum
For stem cell applications where nutrient-rich FBS is required, we now offer mesenchymal stem cell qualified fetal bovine serum (ES-020-B). To reduce lot-to-lot variability, each lot of FBS is extensively screened and qualified to support human MSC cultures using a rigorous clonal growth assay.
Mesenchymal Stem Cell Differentiation Media
Ordering Info
Osteocyte Adipocyte Chondrocyte
Mesenchymal Stem Cell Osteogenic Differentiation Medium (PromoCell)*
Mesenchymal Stem Cell Adipogenic Differentiation Medium 2 (PromoCell)*
Mesenchymal Stem Cell Chondrogenic Differentiation Medium (PromoCell)*
OsteoMAX-XF Osteogenesis Differentiation Medium (Sigma) AdipoMAX Adipogenesis Differentiation Medium (Sigma) ChondroMAX Chondrogenesis Differentiation Medium (Sigma)
*Application Note: Trilineage Differentiation of Multipotent Human Mesenchymal Stem Cells (MSCs) into Osteocytes, Adipocytes and Chondrocytes.
Product Focus: OsteoMAX-XF™ Differentiation Medium
OsteoMAX-XF™ Differentiation Medium is specially formulated to readily differentiate human mesenchymal stem cells into mature osteocytes as assessed by alizarin red and alkaline phosphatase staining. This proprietary formulation was developed using Plasticell’s CombiCult™, a patented, bead-based, combinatorial technology specially developed for discovery of novel stem cell differentiation protocols.
Learn About Combicult Technology
Application Note: Accelerated Osteocyte Differentiation of Human Mesenchymal Stem Cells in a Novel Xeno-free and Serum-free Osteogenesis Differentiation Media
Features and Benefits
  • Serum-free, xeno-free, defined medium formulation
  • Rapid, robust, and highly efficient mineralization and bone nodule formation in standard tissue culture system by as early as 7 days of differentiation.
  • Proven to work in 3D BioPrinting applications
“OsteoMAX-XF produces tissue of impressive osteogenic potential, including key bone-specific and mineralization markers. We found we could rapidly fabricate functional, fully human tissue derived from MSCs and the ECM proteins they produce using the NovoGen MMX 3D-Bioprinter”
Dr. Eric Michael David MD JD, Chief Strategy Officer of Organovo
Mineralization kinetics of human bone-marrow derived MSC
Figure 4. Mineralization kinetics of human bone-marrow derived MSC (Product No. SCC034) differentiated in OsteoMAX-XF™ Medium vs. leading competitor osteogenesis media.
 Mesenchymal Stem Cell Markers
Positive Markers: CD105, CD73, CD90, STRO-1
Negative Markers: CD45, CD34, CD14, CD11b, CD19 and HLA-DR
Browse all of our Mesenchymal Stem Cell Antibodies here
Mesenchymal Stem Cell Related Stains and Dyes
Alizarin Red  |  Oil Red O  |  Safranin  |  Alcian Blue
Human Mesenchymal Stem Cell Characterization Kit
The Human Mesenchymal Stem Cell Characterization Kit contains a panel of validated positive and negative selection markers for the characterization of the mesenchymal stem cell population in human samples. The kit contains positive antibodies directed against CD44, CD90, STRO-1, and CD146. In addition, two specific hematopoietic antibodies CD14 and CD19.
Mesenchymal Stem Cell Markers
Figure 5. Immunocytochemical staining of cultured human bone marrow-derived mesenchymal stem cells stained with antibodies provided in the kit. Nuclei of the cells were visualized with DAPI (blue). CD19 and CD14 staining were not present on human mesenchymal stem cells.
Cell Culture Products and Reagents