[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Applications](https://www.sigmaaldrich.com/US/en/applications)[Materials Science and Engineering](https://www.sigmaaldrich.com/US/en/applications/materials-science-and-engineering) 3D Printing
# 3D Printing
3D printing, also called additive manufacturing, is a process for making a three-dimensional object from a digital model by successively fabricating material layer-by-layer in an additive manner. First, a virtual 3D representation of the object is rendered using computer-aided design (CAD). Then, the model is ‘sliced’ into a series of horizontal layers to convert the design into an STL (standard tessellation language) file that is readable for the 3D printer. This data is then transferred to the printer, and the printer settings are defined. The final object is produced one layer at a time, with each layer bonding and building on the previous one.
Due to the ability to produce very complex shapes and structures with high precision and repeatability from a diverse range of materials, 3D printing is used for a broad range of applications in aerospace, automotive, construction, fashion, food, jewelry, manufacturing, and medical sectors. There are numerous techniques to 3D print objects with different mechanical, thermal, and chemical properties from materials in molten, liquid, or powder states.
* * *
## Related Products
Slide 1 of 20
1 of 5
[
\
Sigma-Aldrich
\
TRD3D0042
\
Flexmark 7 TPU flexible 3D printing filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0042)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0048
\
Flexability TPE-HP flexible3D printing filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0048)
Quick View
[
\
Sigma-Aldrich
\
900983
\
Flexible Thermoplastic Copolyester (TPC) Printing Filament](https://www.sigmaaldrich.com/US/en/product/aldrich/900983)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0025
\
Kyotoflex BioFlexible PLA 3D printing filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0025)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0030
\
Performance ABS 3D Printing Filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0030)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0060
\
Tenax PC-ABS 3D Printing Filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0060)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0055
\
Longchain Nylon 3D Printing Filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0055)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0021
\
Gonzales High Speed PLA 3D Printing Filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0021)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0024
\
Gonzales High Speed PLA 3D Printing Filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0024)
Quick View
[
\
Sigma-Aldrich
\
TRD3D0039
\
Stiron HIPS 3D Printing Filament](https://www.sigmaaldrich.com/US/en/product/aldrich/trd3d0039)
Quick View
[
\
Sigma-Aldrich
\
901029
\
Polyvinyl alcohol (PVA) printing filament](https://www.sigmaaldrich.com/US/en/product/aldrich/901029)
Quick View
[
\
Sigma-Aldrich
\
901031
\
Polyvinyl alcohol (PVA) printing filament](https://www.sigmaaldrich.com/US/en/product/aldrich/901031)
Quick View
[
\
Sigma-Aldrich
\
808156
\
3D printing graphene ink](https://www.sigmaaldrich.com/US/en/product/aldrich/808156)
Quick View
[
\
Sigma-Aldrich
\
901662
\
3D Printing Graphite Ink](https://www.sigmaaldrich.com/US/en/product/aldrich/901662)
Quick View
[
\
Sigma-Aldrich
\
902950
\
Conductive ink for solvent-cast 3D printers](https://www.sigmaaldrich.com/US/en/product/aldrich/902950)
Quick View
[
\
Sigma-Aldrich
\
902969
\
Conductive ink for solvent-cast 3D printers](https://www.sigmaaldrich.com/US/en/product/aldrich/902969)
Quick View
[
\
Sigma-Aldrich
\
902977
\
Conductive ink for solvent-cast 3D printers](https://www.sigmaaldrich.com/US/en/product/aldrich/902977)
Quick View
[
\
Sigma-Aldrich
\
901889
\
3D printing copper ink](https://www.sigmaaldrich.com/US/en/product/aldrich/901889)
Quick View
[
\
Sigma-Aldrich
\
901886
\
3D Printing Zirconia ink](https://www.sigmaaldrich.com/US/en/product/aldrich/901886)
Quick View
[
\
Sigma-Aldrich
\
902403
\
3D Printing Hydroxyapatite ink](https://www.sigmaaldrich.com/US/en/product/aldrich/902403)
Quick View
* * *
## Featured Categories
[](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
[Oxides & Ceramics](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
We offer an extensive range of high purity oxides and ceramics with specialized synthesis and purification techniques, and distinct particle sizes.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
[](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
[Carbon Nanomaterials](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
Discover diverse carbon nanomaterials: fullerene, nanotubes, graphene, quantum dots, nanodiamonds. Fuel energy, electronics, therapeutics research.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
[](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
[Metals](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
Metals of Excellence: Elevate R&D, production, and quality control with our varied metal offerings. Unmatched purity and batch-to-batch consistency.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
[](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
[Specialty & Smart Polymers](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
Find specialized polymers for high-strength structural applications, including thermoset, thermoplastic, and 3D printing options.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
* * *
Overview
Related Articles & Protocols
Support
## Vat Polymerization
Vat polymerization uses photopolymerization to cure and solidify liquid polymer resin. Stereolithography (SL) was the first type of 3D printing developed and commercialized using this method. An SL printer uses mirrors positioned at the X-Y axes to direct a laser beam across a vat of resin to create a cross-section of the object. Digital light processing (DLP) uses a projector with an LCD screen or UV light source to flash light, creating each layer. This type of resin-based 3D printing is fast because an entire layer is exposed all at once. Masked stereolithography (MLA) uses an array of LEDs to shine UV light through a liquid crystal display (LCD) photomask.
## Fused Deposition Modeling(FDM)
Fused deposition modeling (FDM), also called fused filament fabrication (FFF) or material extrusion, is the most common and inexpensive 3D printing technology. A spool of thermoplastic filament (e.g., PLA, ABS) is heated to its melting point and extruded through a nozzle onto a platform, where the molten material cools and solidifies. This technology is used in injection molding and modern plastic manufacturing for ready-to-use products.
## Powder Bed Fusion
Powder bed fusion selectively cures polymer or metal powders with a thermal energy source to create a solid plastic or metal object. First, the powder is heated to a temperature just below its melting point. Then, a roller distributes a very thin layer of powder over the surface of the building bed before a laser passes over the layer to fuse it. Once a layer is completed, the powder bed sinks incrementally to form the next layer. Selective laser sintering (SLS) successively sinters polymer powder with a laser. Selective laser melting (SLM) involves fully melting metal powder instead of sintering it. Other forms of metal powder bed fusion include direct metal laser sintering (DMLS) and electron beam melting (EBM).
## Jetting
Material jetting uses inkjet printer technology to add tiny droplets of photopolymers or wax onto a build plate. An ultraviolet (UV) light simultaneously cures the layers as they are printed. Material jetting (MJ) deposits material in a rapid, line-wise fashion, rather than point-wise. Therefore, multiple objects can be fabricated in a single line. Moreover, this method allows different materials to be printed in the same object. Drop-on-demand (DOD) 3D printing technology uses a set of two inkjets to deposit both the final object material and dissolvable support material simultaneously.
[](https://www.sigmaaldrich.com/documents-search)
[Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search)
Visit our document search for data sheets, certificates and technical documentation.
[Find Documents](https://www.sigmaaldrich.com/documents-search)
[Shop All Products](https://www.sigmaaldrich.com/search/titration?focus=products&page=1&perpage=30&sort=relevance&term=Titration&type=product)
Slide 1 of 6
__Binder Jetting 3D Printing Process__
__Material Jetting 3D Printing Process__
__Power Bed Fusion 3D Printing Process__
 3D Printing Process")
__Digital Light Processing (DLP) 3D Printing Process__
__Stereolithography Printing Process__
 Printing Process")
__Fused Deposition Modeling (FDM) Printing Process__
## Related Articles
- [3D Printable Inks: Energy & Environmental Apps](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-printable-inks-for-energy-and-environmental-applications)
Additive manufacturing combined with nanomaterial-based inks enhances material properties, particularly in energy and environmental applications.
- [3D Printable Conductive Nanocomposites of PLA and Multi-walled Carbon Nanotubes](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-printable-conductive-nanocomposites)
A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.
- [3D/4D Printing Technologies](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-and-4d-printing-technologies)
Three-dimensional printing technology applications range from personal tools to aerospace equipment.
- [Graphene Inks for Electronics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/graphene-inks-for-printed-electronics)
Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.
- [Inkjet Printing for Printed Electronics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/inkjet-printing)
Inkjet printing is one of the key enabling technologies of printed electronics. Inkjet printing technology classification, aspects of materials (inks, substrates) and respective pre-and post-processing steps are discussed.
- [See All (19)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
## Related Protocols
- [Characterization and Performance Insights of Advanced Li-ion Cathode Materials](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/characterization-performance-insights-advanced-li-ion-cathode-materials)
Analysis of high- to moderate-nickel cathode materials for lithium-ion batteries, focusing on performance, stability, and structural characteristics.
- [Streamlined Electrode Fabrication: A User-Friendly Guide](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/streamlined-electrode-fabrication-user-friendly-guide-cathode-electrodes)
Guide for fabricating NMC811 cathode electrodes, detailing procedures for optimal performance in lithium-ion batteries.
- [Preparation of Monodisperse Polymer Spheres](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/nanoparticle-and-microparticle-synthesis/preparation-of-monodisperse-polymer-spheres)
Polymeric spheres serve as crystal templates. Synthesis methods yield large quantities.
- [See All (3)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
### Find More Articles and Protocols
Enter Keywords
Search
## How Can We Help
In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/US/en/support/customer-support)
or talk to our customer service team:
Email [custserv@sial.com](mailto:custserv@sial.com)
or call +1 (800) 244-1173
## Additional Support
- [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search)
Use the Chromatogram Search to identify unknown compounds in your sample.
- [Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- [Customer Support Request](https://www.sigmaaldrich.com/US/en/support/customer-support)
Customer support including help with orders, products, accounts, and website technical issues.
- [FAQ](https://maestro.my.site.com/knowledgeportal/s/)
Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services.
* * *
__Featured Articles__
[3D Printable Inks: Energy & Environmental Apps](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-printable-inks-for-energy-and-environmental-applications)
Additive manufacturing combined with nanomaterial-based inks enhances material properties, particularly in energy and environmental applications.
[3D Printable Conductive Nanocomposites of PLA and Multi-walled Carbon Nanotubes](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-printable-conductive-nanocomposites)
A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.
[3D/4D Printing Technologies](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-and-4d-printing-technologies)
Three-dimensional printing technology applications range from personal tools to aerospace equipment.
[Graphene Inks for Electronics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/graphene-inks-for-printed-electronics)
Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)