[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Applications](https://www.sigmaaldrich.com/US/en/applications)[Materials Science and Engineering](https://www.sigmaaldrich.com/US/en/applications/materials-science-and-engineering) Photovoltaics & Solar Cells
# Photovoltaics & Solar Cells
Solar photovoltaic technology converts light into electricity at the atomic level. The photoelectric effect causes certain semiconductor materials to absorb sunlight particles or photons, and release electrons. A photovoltaic cell produces electricity from visible light; a solar cell absorbs the full range of light frequencies, not just visible light, from sunlight and converts solar radiation into useful energy. As a safe, sustainable, and efficient source of energy, photovoltaic and solar cell systems are used for network or isolated power generation in many types of devices, from electric vehicles (EVs) and solar roofs to water pumping and desalination systems.
Photovoltaic cells utilize layered semiconductor materials as a PN junction to convert light energy in the form of photons to electric current in the form of electrons. The PN junction is an interface between a p-type semiconductor (electron acceptor material) and an n-type semiconductor (electron donor material). When the photon is absorbed by the n-type semiconductor, an electron is dislodged, generating a free electron and electron-hole pair. The negatively charged electron is attracted to the p-type material, and the positively charged hole is attracted to the n-type material. If a completed circuit is connected to the electrodes, the free electron will travel through the circuit, creating electric current and voltage, until it recombines with an electron-hole back in the p-type material.
* * *
## Related Products
Slide 1 of 20
1 of 5
[
\
Sigma-Aldrich
\
900168
\
Lead(II) iodide](https://www.sigmaaldrich.com/US/en/product/aldrich/900168)
Quick View
[
\
Sigma-Aldrich
\
808350
\
TokeOni](https://www.sigmaaldrich.com/US/en/product/aldrich/808350)
Quick View
[
\
Sigma-Aldrich
\
792071
\
Spiro-MeOTAD](https://www.sigmaaldrich.com/US/en/product/aldrich/792071)
Quick View
[
\
Sigma-Aldrich
\
554359
\
Lead(II) iodide](https://www.sigmaaldrich.com/US/en/product/aldrich/554359)
Quick View
[
\
Sigma-Aldrich
\
431648
\
Sodium sulfide nonahydrate](https://www.sigmaaldrich.com/US/en/product/aldrich/431648)
Quick View
[
\
Sigma-Aldrich
\
409308
\
Tin(II) iodide](https://www.sigmaaldrich.com/US/en/product/aldrich/409308)
Quick View
[
\
Sigma-Aldrich
\
229393
\
Aluminum chloride hydrate](https://www.sigmaaldrich.com/US/en/product/aldrich/229393)
Quick View
[
\
Sigma-Aldrich
\
206237
\
Iridium(IV) oxide](https://www.sigmaaldrich.com/US/en/product/aldrich/206237)
Quick View
[
\
Sigma-Aldrich
\
202908
\
Cadmium chloride](https://www.sigmaaldrich.com/US/en/product/aldrich/202908)
Quick View
[
\
Sigma-Aldrich
\
288497
\
Barium oxide](https://www.sigmaaldrich.com/US/en/product/aldrich/288497)
Quick View
[
\
Sigma-Aldrich
\
345865
\
Ammonium trifluoromethanesulfonate](https://www.sigmaaldrich.com/US/en/product/aldrich/345865)
Quick View
[
\
Sigma-Aldrich
\
258385
\
Protoporphyrin IX disodium salt](https://www.sigmaaldrich.com/US/en/product/aldrich/258385)
Quick View
[
\
Sigma-Aldrich
\
637238
\
Silicon dioxide](https://www.sigmaaldrich.com/US/en/product/aldrich/637238)
Quick View
[
\
Sigma-Aldrich
\
902500
\
SHT-263 Solarpur®](https://www.sigmaaldrich.com/US/en/product/aldrich/902500)
Quick View
[
\
Sigma-Aldrich
\
718467
\
Titanium(IV) oxide](https://www.sigmaaldrich.com/US/en/product/aldrich/718467)
Quick View
[
\
Sigma-Aldrich
\
666610
\
Tetrakis(dimethylamido)hafnium(IV)](https://www.sigmaaldrich.com/US/en/product/aldrich/666610)
Quick View
[
\
Sigma-Aldrich
\
718475
\
Aluminum oxide](https://www.sigmaaldrich.com/US/en/product/aldrich/718475)
Quick View
[
\
Sigma-Aldrich
\
425311
\
IR-780 iodide](https://www.sigmaaldrich.com/US/en/product/aldrich/425311)
Quick View
[
\
Sigma-Aldrich
\
333972
\
Zirconium(IV) propoxide solution](https://www.sigmaaldrich.com/US/en/product/aldrich/333972)
Quick View
[
\
Sigma-Aldrich
\
901434
\
Methylammonium iodide](https://www.sigmaaldrich.com/US/en/product/aldrich/901434)
Quick View
* * *
## Featured Categories
[](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/solar-energy-materials)
[Solar Energy Materials](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/solar-energy-materials)
We offer powerful and stable solar energy materials, perovskites, quantum dots, and lead-free alternatives to increase the power conversion efficiency of your renewable energy applications.
[View Products](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/solar-energy-materials)
[](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/high-purity-salts)
[High-Purity Salts](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/high-purity-salts)
We offer a wide range of high-purity salts, including nitrates, oxalates, halides, sulfates, carbonates, and acetates, available in both anhydrous and hydrated forms.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/high-purity-salts)
[](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/solution-and-vapor-deposition-precursors)
[Solution & Vapor Deposition Precursors](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/solution-and-vapor-deposition-precursors)
Our high-quality solution deposition and vapor deposition precursors are optimal for...
[View Products](https://www.sigmaaldrich.com/US/en/products/materials-science/energy-materials/solution-and-vapor-deposition-precursors)
[](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
[Inorganic & Metallic Nanomaterials](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
We offer a comprehensive portfolio of inorganic and metallic nanomaterials, functionalized nanoparticles, and nanomaterial kits for your research needs.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
* * *
Overview
Related Articles & Protocols
Support
The efficiency of photovoltaic systems vary by the type of photovoltaic cell technology and the type of semiconductor material used. The first solar cells were composed of inorganic polycrystalline and single-crystalline materials. Notable progress has been made in photovoltaic technology due to remarkable advancements in organic electronics and materials.
An organic solar cell is lightweight, flexible and can be produced at low cost with high-performance polymeric donors, fullerene, and non-fullerene acceptors (NFAs) through low-temperature solution processes on a transparent conductor, such as indium tin oxide (ITO) or fluorine-doped tin oxide (FTO). Organic hole-transport materials (HTMs) have enabled high-performance perovskite solar cells as an alternative, more efficient method for harvesting solar energy.
Perovskite solar cells typically use a hybrid inorganic-organic material as the light-harvesting active layer. Perovskite solar cells benefit from a high conversion efficiency, low cost, and simple manufacturing, making them the fastest advancing solar technologies for commercial applications. Lead halide perovskites have the highest conversion efficiency and are the fastest growing solar cell technology.
[](https://www.sigmaaldrich.com/documents-search)
[Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search)
Visit our document search for data sheets, certificates and technical documentation.
[Find Documents](https://www.sigmaaldrich.com/documents-search)
## Related Articles
- [Metal Organic Frameworks (MOFs)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/metal-organic-frameworks)
Metal-organic frameworks (MOFs) are porous, crystalline materials. MOFs are composed of metal ions or clusters bound by multidirectional organic ligands, which act as linkers in the network structure.
- [Ruthenium-based dyes for Dye-sensitized Solar Cells](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/dye-solar-cells)
Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.
- [Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/synthesis-properties)
Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures
- [Quantum Dots for Electronics and Energy Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/quantum-dots-noncadmium)
In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.
- [Redi Dri™ Packaging for Consistent Free-Flow in Hygroscopic Salts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/redi-dri-packaging-consistent-flow-hygroscopic-salts)
Achieve moisture control and stability in hygroscopic salts with Redi-Dri™ packaging for consistent flow, purity, and trace metal stability.
- [See All (39)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
## Related Protocols
- [Characterization and Performance Insights of Advanced Li-ion Cathode Materials](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/characterization-performance-insights-advanced-li-ion-cathode-materials)
Analysis of high- to moderate-nickel cathode materials for lithium-ion batteries, focusing on performance, stability, and structural characteristics.
- [Streamlined Electrode Fabrication: A User-Friendly Guide](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/streamlined-electrode-fabrication-user-friendly-guide-cathode-electrodes)
Guide for fabricating NMC811 cathode electrodes, detailing procedures for optimal performance in lithium-ion batteries.
- [Preparation of Monodisperse Polymer Spheres](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/nanoparticle-and-microparticle-synthesis/preparation-of-monodisperse-polymer-spheres)
Polymeric spheres serve as crystal templates. Synthesis methods yield large quantities.
- [See All (3)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
### Find More Articles and Protocols
Enter Keywords
Search
## How Can We Help
In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/US/en/support/customer-support)
or talk to our customer service team:
Email [custserv@sial.com](mailto:custserv@sial.com)
or call +1 (800) 244-1173
## Additional Support
- [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search)
Use the Chromatogram Search to identify unknown compounds in your sample.
- [Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- [Customer Support Request](https://www.sigmaaldrich.com/US/en/support/customer-support)
Customer support including help with orders, products, accounts, and website technical issues.
- [FAQ](https://maestro.my.site.com/knowledgeportal/s/)
Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services.
* * *
__Featured Articles__
[Metal Organic Frameworks (MOFs)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/metal-organic-frameworks)
Metal-organic frameworks (MOFs) are porous, crystalline materials. MOFs are composed of metal ions or clusters bound by multidirectional organic ligands, which act as linkers in the network structure.
[Ruthenium-based dyes for Dye-sensitized Solar Cells](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/dye-solar-cells)
Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.
[Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/synthesis-properties)
Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures
[Quantum Dots for Electronics and Energy Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/photovoltaics-and-solar-cells/quantum-dots-noncadmium)
In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.