Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
powder
concentration
<34.8% (by Na2S2O3, titration)
SMILES string
O.[Mg++].[O-][Cr]([O-])(=O)=O
InChI
1S/Cr.Mg.H2O.4O/h;;1H2;;;;/q;+2;;;;2*-1
InChI key
BWDANDPWLRGULC-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
23371-94-0
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


