Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C2H14B2N2
CAS Number:
Molecular Weight:
87.77
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23
Assay:
96%
Form:
solid
assay
96%
form
solid
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
114-130 °C (dec.)
application(s)
hydrogen and fuel cell
greener alternative category
SMILES string
B.B.NCCN
InChI
1S/C2H8N2.2BH3/c3-1-2-4;;/h1-4H2;2*1H3
InChI key
MZYKPEFBHALNHU-UHFFFAOYSA-N
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Application
Enantioselective hydride attack of borane-ethylenediamine complex on inclusion compounds of ketones and certain diols and β-cyclodextrin may form optically active alcohols.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
4.1B - Flammable solid hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

