Skip to Content

799246

Poly(L-lactide-block-acrylic acid)

PLLA average Mn 10,000, PAA average Mn 8,000

Synonym(s):

PLLA-block-PAA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H25S3(C3H4O2)yC6H9NO(C3H4O2)xH
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

mol wt

PAA average Mn 8,000, PLLA average Mn 10,000

storage temp.

−20°C

Application

The PLLA-block-PAA is a biocompatible diblock copolymer which forms cylindrical micelles. The PLLA is hydrophobic and a degradable block and the PAA is hydrophilic.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library