Skip to Content
Merck

802174

Poly(L-lactide) 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentonate

average Mn 18,000, PDI ≤1.2

Synonym(s):

PLA RAFT, PLA thiocarbonyl, PLLA RAFT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C19H35NOS3(C3H4O2)n
UNSPSC Code:
12162002
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

mol wt

average Mn 18,000

PDI

≤1.2

storage temp.

2-8°C

Application

The Poly(L-lactide) is a biodegradable polymer that contains RAFT agent functionality. The benzodithioate moiety is well suited to polymerize methacrylates and methacrylamides. PLA is commonly used in several applications, including Drug Delivery, nanoparticle surface modification, end-group functionalization, macroinitiator or block precursor.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents: WO98/01478; WO99/311444.


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library