Skip to Content
Merck

Skip To

GF08074650

Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8

foil, 0.5m coil, 0.5mm thickness, annealed

Synonym(s):

NI020400

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.23
UNSPSC Code:
11101712
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

annealed, foil (coil)

composition

Ni72Cr16Fe8

reaction suitability

core: chromium, core: iron, core: nickel

manufacturer/tradename

Goodfellow 080-746-50

size × thickness

0.5 m × 0.5 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF59961572GF23075681GF69539271
form

annealed, foil (coil)

form

annealed, foil (coil)

form

annealed, foil (coil)

form

annealed, foil (coil)

manufacturer/tradename

Goodfellow 080-746-50

manufacturer/tradename

Goodfellow 599-615-72

manufacturer/tradename

Goodfellow 230-756-81

manufacturer/tradename

Goodfellow 695-392-71

size × thickness

0.5 m × 0.5 mm

size × thickness

0.2 m × 0.5 mm

size × thickness

2 m × 0.1 mm

size × thickness

0.5 m × 0.075 mm

reaction suitability

core: chromium, core: nickel

reaction suitability

core: chromium, core: iron, core: nickel

reaction suitability

core: chromium, core: iron, core: nickel

reaction suitability

core: chromium, core: iron, core: nickel


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters