Skip to Content
Merck

Skip To

GF26067775

Aluminum

tube, outside diameter 12.0 mm, length 200 mm, wall thickness 0.5 mm

Synonym(s):

AL007500

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
$536.00
5 ea
Please contact Customer Service for Availability
$2,170.00
10 ea
Please contact Customer Service for Availability
$3,970.00

About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141702
MDL number:

$536.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tubes

autoignition temp.

1400 °F

manufacturer/tradename

Goodfellow 260-677-75

resistivity

2.6548 μΩ-cm

L

200 mm

O.D.

12.0 mm

wall thickness

0.5 mm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF49286201GF99970082GF67101308
form

tubes

form

tubes

form

tubes

form

tubes

manufacturer/tradename

Goodfellow 260-677-75

manufacturer/tradename

Goodfellow 492-862-01

manufacturer/tradename

Goodfellow 999-700-82

manufacturer/tradename

Goodfellow 671-013-08

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

mp

660.37 °C (lit.)

mp

660.37 °C (lit.)

mp

660.37 °C (lit.)

mp

660.37 °C (lit.)


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters