Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.23
UNSPSC Code:
12352100
form
film
manufacturer/tradename
Goodfellow 353-662-49
L
0.5 m
coil width
63.5 mm
thickness
0.76 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Still not finding the right product?
Explore all of our products under Polyaramid
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.