Skip to Content
Merck

GF39699837

High permeability magnetic alloy, Ni80Fe15Mo5

foil, 6mm disks, 0.0075mm thickness

Synonym(s):

NI180210

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352300
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

foil (disc)

manufacturer/tradename

Goodfellow 396-998-37

diam. × thickness

6 mm × 0.0075 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library