Skip to Content

GF50833009

Aluminum

foil, thickness 1.0 mm, 25 mm diameter, purity 99%

Synonym(s):

AL000700

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Al
CAS Number:
Molecular Weight:
26.98
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141702
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

form

foil

autoignition temp.

1400 °F

manufacturer/tradename

Goodfellow 508-330-09

resistivity

2.6548 μΩ-cm

diam.

25 mm

thickness

1.0 mm

diameter

25 mm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library