Skip to Content
Merck

Skip To

GF99119986

Alumina

sheet, thickness 5.0 mm, size 25 × 25 mm, purity 99%

Synonym(s):

AL603450

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.23
UNSPSC Code:
11181511
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

form

sheet

manufacturer/tradename

Goodfellow 991-199-86

size

25 × 25 mm

thickness

5.0 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF89998159GF15192952GF49768880
assay

99%

assay

99%

assay

99%

assay

99%

form

sheet

form

sheet

form

sheet

form

sheet

manufacturer/tradename

Goodfellow 991-199-86

manufacturer/tradename

Goodfellow 899-981-59

manufacturer/tradename

Goodfellow 151-929-52

manufacturer/tradename

Goodfellow 497-688-80

thickness

5.0 mm

thickness

5.0 mm

thickness

4.0 mm

thickness

5.0 mm

size

25 × 25 mm

size

50 × 50 mm

size

25 × 50 mm

size

100 × 100 mm

99.99% alumina for laboratory use

ACS grade alumina for laboratory use, Al2O3 alumina for research purposes, activated alumina for adsorbent applications, alumina catalyst support material, alumina ceramic material, alumina desiccant, alumina for dental ceramics, alumina for industrial applications, alumina nanoparticles for coatings, alumina powder, alumina substrate for semiconductor fabrication, calcined alumina for refractory applications, high purity alumina 99.99%, lab-grade alumina for chromatography

-

-


Still not finding the right product?

Explore all of our products under Alumina




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters