Skip to Content

H-180

6-Hydroxy Buspirone solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H31N5O3
CAS Number:
Molecular Weight:
401.50
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O=C1C(O)C2(CCCC2)CC(N1CCCCN(CC3)CCN3C4=NC=CC=N4)=O

InChI

1S/C21H31N5O3/c27-17-16-21(6-1-2-7-21)18(28)19(29)26(17)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20/h5,8-9,18,28H,1-4,6-7,10-16H2

InChI key

KOZNAHJIJGCFJJ-UHFFFAOYSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F

flash_point_c

2 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library