Skip to Content

03-182

HDAC2 (1-488) (His-tag) human recombinant

Human recombinant HDAC2 (1-488) (His-tag) produced in insect cells.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.32
eCl@ss:
32160405
Technique(s):
activity assay: suitable
Species reactivity:
-
Application:
activity assay
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

specific activity

>5 units/μg

mol wt

Mw ~55 kDa

manufacturer/tradename

Upstate®

concentration

>50% (SDS-PAGE)

technique(s)

activity assay: suitable

shipped in

ambient

General description

Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDAC2 is likely to be involved in regulating chromatin structure during transcription. It associates with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA. This process is highly conserved from yeast to human.

Application

HDAC deacetylase assays
Human recombinant HDAC2 (1-488) (His-tag) produced in insect cells.
Research Category
Epigenetics & Nuclear Function

Physical form

Format: Purified
In liquid containing 50 mM Tris, pH 8.0, 138 mM NaCl and 10% glycerol. Frozen solution.

Preparation Note

Store at -80°C. The enzyme is stable on ice for 30 to 60 min, but may lose activity with dilution and/or prolonged storage on ice. It is recommended that thawing and addition of the enzyme to assay wells be done within as short a time as possible before start of the assay and that the thawed enzyme be kept on ice throughout. Best performance will be obtained by adding the chilled, undiluted enzyme directly to pre-warmed buffer and proceeding immediately to the addition of pre-warmed substrate. The remaining, unused enzyme should be refrozen quickly by, for example, snap freezing in a dry/ice ethanol bath or liquid nitrogen. The enzyme is stable to at least 4 freeze/thaw cycles. To minimize the number of freeze/thaw cycles, aliquot the HDAC2 into separate tubes before storing at -80°C.

Other Notes

Specific Activity: 1U = 1 pmol/min at 37°C using a standard flourometric activity assay to measure deacetylation activity.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library