Skip to Content

8.06715

Nickel(II) acetylacetonate

for synthesis

Synonym(s):

Nickel(II) acetylacetonate, Bis(acetylacetonato)nickel(II), Bis(2,4-pentanedionato) nickel(II)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H14NiO4
CAS Number:
Molecular Weight:
256.91
EC Index Number:
221-875-7
UNSPSC Code:
12352103
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

2.7 hPa ( 110 °C)

Quality Segment

form

powder

potency

587 mg/kg LD50, oral (Rat)

mp

230 °C

solubility

4.8 g/L

bulk density

470 kg/m3

storage temp.

2-30°C

InChI

1S/2C5H8O2.Ni/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;

InChI key

SHWZFQPXYGHRKT-UHFFFAOYSA-N

Analysis Note

Assay (Ni): ≥ 98,0 %(m)
Identity (IR): conforms


signalword

Danger

flash_point_f

428.0 °F

flash_point_c

220 °C

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library