Skip to Content

0720-0020

Seraseq Euploid Rm (12% Target Fetal Fraction)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

description

Seraseq® Reference Material

manufacturer/tradename

LGC

technique(s)

DNA sequencing: suitable (NGS)

application(s)

clinical testing
genomic analysis (NGS)

storage temp.

2-8°C

General description

A full-process negative-control NIPT reference material for NGS-based detection of common fetal trisomies in cell-free maternal DNA.
This product is provided as delivered and specified by the manufacturer. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued by the manufacturer. For further information and support, please go to the website of the manufacturer at https://www.seracare.com/Controls-Reference-Materials

Analysis Note

These products are for research use only and not for use in diagnostic procedures.

Other Notes

Sales restrictions may apply.
Seraseq® is a registered trademark of LGC Clinical Diagnostics, Inc. © 2024 LGC Clinical Diagnostics, Inc. All rights reserved. LGC issued SDS can be found at https://www.seracare.com/Controls-Reference-Materials

Legal Information

Seraseq is a registered trademark of LGC Clinical Diagnostics, Inc.


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library