Skip to Content

11370

8-Azaadenine

≥99.0% (HPLC)

Synonym(s):

6-Amino-8-azapurine, 8-Aza-6-aminopurine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H4N6
CAS Number:
Molecular Weight:
136.11
EC Number:
214-375-5
UNSPSC Code:
12352202
MDL number:
Beilstein/REAXYS Number:
141551
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (HPLC)

mp

>300 °C (lit.)

SMILES string

Nc1ncnc2nn[nH]c12

InChI

1S/C4H4N6/c5-3-2-4(7-1-6-3)9-10-8-2/h1H,(H3,5,6,7,8,9,10)

InChI key

HRYKDUPGBWLLHO-UHFFFAOYSA-N

General description

Base pairs of azaadenine with thymine (and azaguanine with cytosine) are found by ab initio calculation to be up to 7 kcal/mol more stable than natural AT (and GC) base pairs.

Other Notes

Inhibits germination of Cicer arientium seeds, a system used to stimulate embryonic tissue




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library