Skip to Content

32632

Dimethachlor Metabolite CGA 373464

PESTANAL®, analytical standard

Synonym(s):

[(2,6-Dimethylphenyl)(2-sulfoacetyl)amino]acetic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H14NNaO6S
CAS Number:
Molecular Weight:
323.30
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na+].Cc1cccc(C)c1N(CC(O)=O)C(=O)CS([O-])(=O)=O

InChI

1S/C12H15NO6S.Na/c1-8-4-3-5-9(2)12(8)13(6-11(15)16)10(14)7-20(17,18)19;/h3-5H,6-7H2,1-2H3,(H,15,16)(H,17,18,19);/q;+1/p-1

InChI key

BSGUBTIVHBTTNF-UHFFFAOYSA-M

General description

Dimethachlor Metabolite CGA 373464 is a metabolite of dimethachlor which is considered as a potential contaminant in groundwater.

Application

Dimethachlor Metabolite CGA 373464 can be utilized as an analytical reference standard for the quantification of the analyte in surface water and groundwater samples using direct injection liquid chromatography-electrospray ionization-mass spectrometry (LC-ESI-MS) operating in multiple reaction monitoring (MRM) mode.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library