Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
85151701
grade
certified reference material
agency
BCR®
manufacturer/tradename
JRC
application(s)
cosmetics
food and beverages
format
matrix material
storage temp.
−20°C
Analysis Note
A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage
please visit the JRC webpage
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Antibiotics: Chlortetracycline
Matrix Group: Meat
Antibiotics: Chlortetracycline
Matrix Group: Meat
Legal Information
BCR is a registered trademark of European Commission
Storage Class
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.