Skip to Content
Merck

RE-PV1-1

Recombinant Pis v 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

assay

>95%

mol wt

(16kD)

manufacturer/tradename

InBio

format

neat

storage temp.

−20°C

General description

Source:
E. coli Purification:

Multi-step chromatography.

Formulation: Preservative and carrier-free in 1X PBS, 0.3M NaCl, 0.4M L-Arginine, pH 8.0. Filtered through 0.22µm filter.
Notes:
1.) rPis v 1 has a N-terminal 6xHis-tag

Disclaimer

For research and commercial use in vitro: not for human in vivo or therapeutic use.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library