Skip to Content

30079

Alcalase® CLEA

≥5 U/g

Synonym(s):

Protease from Bacillus licheniformis, cross-linked enzyme aggregate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

specific activity

≥5 U/g

storage temp.

2-8°C

General description

Alcalase®, the commercial form of subtilisin, is obtained from Bacillus licheniformis. It is an endopeptidase produced through submerged fermentation. It is mainly used as a detergent additive. Alcalase has an esterase activity for peptide and amino acid esters.

Other Notes

1 U corresponds to the amount of enzyme which sets free 1 μmol folin-positive amino acids and peptides (as tyrosine) per minute at pH 8.0 and 37°C (casein as substrate)

Legal Information

Alcalase is a registered trademark of Novozymes Corp.


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library