Skip to Content

C7659

Monoclonal Anti-Chorionic gonadotropin (β-subunit) (βHCG) antibody produced in mouse

clone PC-2, diluted ascites fluid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

diluted ascites fluid

clone

PC-2, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, radioimmunoassay: 1:8,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CGB(1082)

Immunogen

HCG

Biochem/physiol Actions

The antibody is specific for the β subunit of HCG. Typical cross-reactivities observed with LH, FSH, and TSH are less than 0.3% in RIA.

Preparation Note

Dilute with buffer: 0.01 M phosphate, pH 7.8, 0.15 M sodium chloride, 0.5% bovine serum albumin and 15 mM sodium azide.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library