Skip to Content

M3172

Mitogen activated protein kinase from rat

≥95% (SDS-PAGE), buffered aqueous glycerol solution, recombinant, expressed in E. coli (N-terminal histidine tagged)

Synonym(s):

ERK2, Extracellular-signal regulated kinase, MAP Kinase Activated from rat, MAPK

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli (N-terminal histidine tagged)

assay

≥95% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

≥500 U/mg

mol wt

42 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

rat ... Mapk1(116590)

Biochem/physiol Actions

MAP kinase is a serine/threonine kinase that is activated by MEK through a phosphorylation cascade. The cascade starts with extracellular signals transmitted by growth factor-regulated receptor tyrosine kinases or G-protein coupled receptors. Dually phosphorylated (Thr and Tyr) MAP kinase translocates from the cytoplasm to the nucleus, where it phosphorylates protein transcription factors, transducing extracellular signals into genomic responses, proliferation or differentiation.
MAP kinase is a serine/threonine kinase that is activated by MEK through a phosphorylation cascade.

Physical form

Solution in 20 mM Tris, pH 7.5, 150 mM NaCl, 1 mM EGTA, 10% glycerol, 1 mM DTT, and 0.03% Brij®.

Other Notes

One unit of activated MAP kinase will transfer 1 nmole of phosphate from ATP to myelin basic protein per min at 30 °C.

Legal Information

Brij is a registered trademark of Croda International PLC


Storage Class

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library