Skip to Content

CLS3789A

Corning® 96-Well Plate

96-well, White, Round Bottom, Polystyrene, Not Treated Microplate without Lid, Nonsterile, 25 per Bag, 100 per case

Synonym(s):

96 well plate, cell culture plate, microplate, plate, spheroid plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122102
NACRES:
NB.15
Material:
round wells, white polystyrene
Sterility:
non-sterile
Binding type:
(not treated)
Feature:
lid: no
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

round wells, white polystyrene

sterility

non-sterile

feature

lid: no

packaging

case of 100, pack of 25

manufacturer/tradename

Corning 3789A

plate wells

330 μL

plate working volume

75 μL (200 μL)

binding type

(not treated)

General description

  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface

Application

Spheroid plate

Features and Benefits

  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm²
  • Without lids, lids supplied separately
  • White microplates enhance luminescent signals and have low background luminescence and fluorescence
  • Designed to reduce well-to-well crosstalk
  • Total volume is 330 µL; recommended working volume of 75 to 200 µL
  • Conforms to standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library