Skip to Content
CLS4440
Corning® Elplasia Plate
6 well, Black/Clear, Round Bottom, ULA Microcavity Plate with Lid, Sterile, Individually Packaged
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122102
NACRES:
NB.02
Material:
black polystyrene , round wells
Sterility:
sterile
Binding type:
(ultra-low attachment)
Feature:
lid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
black polystyrene , round wells
sterility
sterile
feature
lid
packaging
case of 5
manufacturer/tradename
Corning 4440
plate wells
16.8 mL
plate working volume
5 mL
binding type
(ultra-low attachment)
General description
The only plates with a simple “plug and play” protocol for scaffold-free, self-assembly spheroid formation at large volumes. Novel well geometry and surface coatings, including Corning Ultra Low Attachment (ULA) featured on the round bottom plates. Corning ULA surface is hydrophilic, biologically inert and non-degradable, which promotes highly reproducible spheroid formation and easy harvesting.
Corning Elplasia round bottom plates optimal for bulk spheroid formation, collection and expansion. Round bottom plates feature Corning Ultra-Low Attachment surface and are available in 6-, 24- and 96-well formats.
Application
Spheroid plate
Features and Benefits
  • Spheroids may be formed and cultured for 21 or more days
  • Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
  • The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
  • Black opaque sidewalls to reduce well-to-well “cross-talk” -Well suited for fluorescent/luminescent assays
  • Available in multiple formats, two well geometries and surface coating options.
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® Elplasia Plate
Choose from one of the most recent versions:
Certificates of Analysis (COA)