About This Item
UNSPSC Code:
41122102
NACRES:
NB.02
Material:
black polystyrene , square wells
Sterility:
sterile
Binding type:
(plasma treated)
Feature:
lid
material
black polystyrene , square wells
sterility
sterile
feature
lid
manufacturer/tradename
Corning 4447
packaging
case of 5 ×
plate wells
110 μL
plate working volume
20 μL (75 μL)
binding type
(plasma treated)
General description
The only plates with a simple “plug and play” protocol for scaffold-free, self-assembly spheroid formation at large volumes. Corning Elplasia square bottom plates feature a surface with optical qualities suited for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters. The Elplasia square plate is well suited for screening programs as it generates a high volume of spheroids per well, allowing for higher signals per well. Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats.
Application
Spheroid plate
Features and Benefits
- Spheroids may be formed and cultured for 21 or more days
- Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
- The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
- Black opaque sidewalls to reduce well-to-well “cross-talk”
- Well suited for luorescent/luminescent assays
- Available in multiple formats, two well geometries and surface coating options.
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® Elplasia Plate
Choose from one of the most recent versions:
Already Own This Product?